96 Well White/Clear Bottom Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10
Thermo Scientific™
96 Well White/Clear Bottom Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10
Color:White, Clear Bottom; Sterility:Non-sterile; Lid:Without Lid; Type: Optical Polymer Base
Have Questions?
Catalog NumberQuantity
265302
Catalog number 265302
Price (USD)
334.00
Case of 30
Price (USD)
334.00
Case of 30
Thermo Scientific™ Nunc MicroWell 96-Well Optical-Bottom Plates with Polymer Base have a solid polystyrene black or white upper structures with a polystyrene film at the bottom of the plate. The plates are optimized for fluorescence and luminescence in cell culture and microscopic applications.
White or black polystyrene upper structure with polymer or coverglass bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications.
  • Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Available in black for fluorescence assays or white for luminescence and fluorescence assays; both with clear bottoms
  • Polymer base plate
  • Working range, 50-200μL/well
  • Customized bar coding is available
Ordering Information:
Part No. 152040, 152036, 152028 and 152037 is produced in a clean environment.
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidWithout Lid
No. per Case30
No. per Pack10
Packaging10 Pk., 30 Cases
Product TypeOptical Polymer Base
Skirt StyleHigh Flange
Volume (Metric) Well400 μL
Volume (Metric) Working50 to 200 μL
Array8 x 12
BarcodeCustom Barcoding Available
ColorWhite
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilityNon-sterile
Surface TreatmentUntreated
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 30
Frequently asked questions (FAQs)
Should I use an opaque black/white immuno plate, or an "optical-bottom" design?
Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.