Trypsin (1:250), powder - FAQs
Trypsin (1:250), powder - FAQs
View additional product information for Trypsin (1:250), powder - FAQs (27250018)
10 product FAQs found
How do I prepare a 1% solution with Trypsin (1:250), powder (Cat. No. 27250018)?
Trypsin (1:250), powder (Cat. No. 27250018) is prepared by adding the 0.25 g of Trypsin powder to 100 mL of chilled HBSS (Cat. No. 14170112) to create a 0.25% solution.
For 1% solution, you would add 1g of trypsin powder to 100 mL of chilled HBSS.
The solution should be mixed for approximately 10 minutes, and the pH should be between 7.4-7.8. The solution should be sterile filtered (0.22 µm filter) and stored -20 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the storage recommendations for reconstituted Trypsin (1:250) and powder (Cat. No. 27250018)?
We recommend storing reconstituted Trypsin (1:250), powder (Cat. No. 27250018) at -20C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the molar concentration of protease in Trypsin (1:250), powder (Cat. No. 27250018)?
When Trypsin (1:250) powder (Cat. No. 27250018) is reconstituted, the protease molarity should be 0.25%. Therefore, the protease molarity should be 0.105 mM.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How do I reconstitute the dry powder for Trypsin (1:250), powder (Cat. No. 27250018)?
We recommend the following instructions to reconstitute Trypsin (1:250), powder:
  1. Add 0.25 g of trypsin powder to 100 mL of chilled HBSS (Cat. No. 14170-112) to create a 0.25% solution.
  2. Mix the solution ~10 minutes and adjust pH to 7.4-7.8.
  3. Filter the solution with a 0.22 µm filter and store at -20°C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
On the Trypsin (1:250), powder (Cat. No. 27250018) product page, what does "1:250" indicate?
1:250 indicates the activity level of the trypsin enzyme. Trypsin 1:250 has the potency to bring about proteolytic digestion of 250 times its weight of casein under conditions specified by the National Formulary.
How long can I keep trypsin in the refrigerator after thawing?
Generally speaking, trypsin can be stored refrigerated for up to a week. However, this may vary depending on how often it is warmed up and used during that week. It may also depend on how tightly the cells are attached, because there will be some loss of activity each day the trypsin is not frozen.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do you have a non-animal-origin enzyme for cell dissociation?
Yes, the TrypLE products contain rProtease, a non-animal, trypsin-like enzyme used for the dissociation of attachment dependent cell lines. TrypLE enzyme has demonstrated the ability to dissociate cells cultured both in serum-free and serum-supplemented systems. The catalog numbers for TrypLE Select are 12563-011 (100 mL) and 12563-029 (500 mL). Some example catalog numbers for TrypLE Express include 12604-013 without Phenol red (100 mL) and 12605-010 with Phenol red (100 mL). Both formulations are also available in additional sizes. All these products are animal origin-free.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between TrypLE Express Enzyme, TrypLE Select Enzyme, and CTS TrypLE Select Enzyme?
All the above products contain rProtease, a non-animal, trypsin-like enzyme formulation used for the dissociation of attachment-dependent cell lines. This alternative for porcine trypsin is a recombinant enzyme derived from microbial fermentation.
The TrpLE Express, TrypLE Select, and CTS TrypLE Select products differ in their production and testing methods and in the documentation provided.
TrypLE Select is for the industrial market and has release-testing both for endotoxins and enzyme activity. This material also has a Drug Master File on record. It is manufactured in AOF-dedicated equipment and is intended for Research Use/Further Cell Culture Manufacturing.
TrypLE Express is manufactured without use of dedicated equipment and is intended for Research Use Only. Not for use in diagnostic procedures.
CTS TrypLE Select is manufactured under scalable cGMP conditions using dedicated equipment and is designed for clinical applications. This product has a Drug Master File on record and is intended for Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. Not intended for direct administration into humans or animals.
To read more about TrypLE and cell release times using this reagent, please go here (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin/tryple-express.html#1).
Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.
What are the benefits of Gibco TrypLE reagent over trypsin?
TrypLE reagent is free of animal- and human-derived components, producing an exceptionally pure reagent that is gentle on cells. Inactivation with trypsin inhibitors is not required. TrypLE reagent is ideal for dissociating a variety of attachment-dependent mammalian cell lines both in serum and in serum-free conditions, and can be directly substituted for trypsin in your current protocol. TrypLE reagent is room-temperature stable and ready to use when you need it. TrypLE cell dissociation reagents remain stable for 24 months at room temperature, making storage and handling easier and more convenient.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What reagents do you offer for cell dissociation, and what are the main differences between them?
Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).
Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.