Nalgene™ Dropper Bottles with Control Dispensing Tip
Nalgene™ Dropper Bottles with Control Dispensing Tip
Nalgene™ Dropper Bottles with Control Dispensing Tip
Nalgene™ Dropper Bottles with Control Dispensing Tip
Nalgene™ Dropper Bottles with Control Dispensing Tip
Nalgene™ Dropper Bottles with Control Dispensing Tip
Thermo Scientific™

Nalgene™ Dropper Bottles with Control Dispensing Tip

Repeatedly dispense reagents accurately using these dropper bottles, an excellent alternative to pipetting devices.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoCapacity (Metric)Closure Color
2752-91254 mLAssorted
2750-91254 mLWhite
2750-90258 mLWhite
2752-90258 mLAssorted
2750-905015 mLWhite
2752-905015 mLAssorted
Número do catálogo 2752-9125
Preço (BRL)
-
Capacity (Metric):
4 mL
Closure Color:
Assorted

Repeatedly dispense reagents accurately using Thermo Scientific™ Nalgene™ Dropper Bottles with Control Dispensing Tip. An excellent alternative to pipetting devices, these dropper bottles deliver 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Featuring excellent chemical resistance, these specialty bottles are suitable for most biotech, diagnostic and pharmaceutical applications.

Features:
  • Translucent bottle becomes contact-clear when liquid is added for easy viewing of contents
  • Dropper control tip snaps into place for a secure fit
  • Dispensing tip and closures included; choose white or multi-colored closures
  • Bottles deliver 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time
  • Leakproof† when used with provided tip and closure/Transparent

Ordering Information:
This product comes in a case of 25.

Recycling Information:
Nalgene reusable LDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.

.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Especificações
Capacity (English)0.13 oz.
Certifications/ComplianceISO 13485:2003
Closure ColorAssorted
Closure Size15-415 mm
For Use With (Equipment)Nalgene Closures
Height (English)2.12 in.
Height (Metric)53.8 mm
No. per Case25
No. per Pack25
Product TypeDropping Bottle
ShapeRound
Through the Closure DispensingYes
Width (English)0.78 in.
Capacity (Metric)4 mL
Closure IncludedYes
ColorTranslucent
Drop Size40 μL
MaterialLDPE
Unit SizeCase of 25