Triton™ X-100 Surfact-Amps™ Detergent Solution, 1 L - FAQs
Triton™ X-100 Surfact-Amps™ Detergent Solution, 1 L - FAQs
View additional product information for Triton™ X-100 Surfact-Amps™ Detergent Solution - FAQs (28313, 85112, 85111, 28314)
2 product FAQs found
What are the storage conditions for Triton X-100 Surfact-Amps Detergent Solution (Cat. No. 85111)?
After opening, we recommend storing the unused solution sealed in an air-tight vial containing minimal air space or from which the air has been purged with nitrogen gas. Frequent opening should be avoided because Triton X-100 is susceptible to autoxidation upon exposure to air.
Can you provide the shelf life for Triton X-100 Surfact-Amps Detergent Solution (Cat. No. 85111)?
Triton X-100 Surfact-Amps Detergent Solution is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (20-25 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).