Brij™-35 Surfact-Amps™ Detergent Solution
Thermo Scientific™

Brij™-35 Surfact-Amps™ Detergent Solution

This Thermo Scientific™ formulation of Brij™ 35 is a stabilized 10% (w/v) solution of Brij-35 detergent for use in variousRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
283166 x 10 mL
85118500 mL
8511750 mL
Catalog number 28316
Price (USD)
155.65
Online exclusive
185.00
Save 29.35
Each
Quantity:
6 x 10 mL
Request bulk or custom format
Price (USD)
155.65
Online exclusive
185.00
Save 29.35
Each
This Thermo Scientific™ formulation of Brij™ 35 is a stabilized 10% (w/v) solution of Brij-35 detergent for use in various protein methods. Brij™ 35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or a surfactant in various HPLC applications.

Properties of Brij 35 Detergent:

• Molecular Weight: 1225g
• Detergent Class: Nonionic
• Aggregation Number: 40
• Micelle Molecular Weight: 49,000g
• Critical Micelle Concentration (CMC): 0.09 mM (0.011%, w/v)
• Cloud Point: >100°C
• Dialyzable: No

Features of this Thermo Scientific detergent solution:

• Brij 35—a nonionic detergent for use in various protein methods
• Accurate—10% detergent solution in ultrapure water
• Easy-to-use—solutions are simple to dispense and dilute for use

Specifications of this Thermo Scientific Brij 35, 10% Solution:

• Visual: Clear, colorless viscous liquid, free of foreign material.
• Concentration: Concentration: 27.0 to 30.0%

Related Products
Brij™-35 Surfact-Amps™ (10% w/v) Solution
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity6 x 10 mL
Reagent TypeDetergent Solution
FormLiquid
Product LineBrij, Surfact-Amps
Product TypeDetergent
Unit SizeEach
Contents & Storage
Store at Room Temperature

Frequently asked questions (FAQs)

What is the Brij 35 detergent concentration in the 1X Zymogram Developing buffer?

The Brij 35 detergent concentration for Zymogram Developing buffer should be 0.02% (w/v).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.