Tween™ 20 Surfact-Amps™ Detergent Solution
Thermo Scientific™

Tween™ 20 Surfact-Amps™ Detergent Solution

Thermo Scientific Tween-20 Surfact-Amps Detergent Solution is a highly-purified Tween™ 20 detergent stabilized as a 10% solution (w/v) and packaged자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
283206 x 10 mL
8511350 mL
85114250 mL
85115500 mL
283211 L
카탈로그 번호 28320
제품 가격(KRW)
213,000
온라인 행사
Ends: 31-Dec-2026
243,000
할인액 30,000 (12%)
Each
수량:
6 x 10 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
213,000
온라인 행사
Ends: 31-Dec-2026
243,000
할인액 30,000 (12%)
Each
Thermo Scientific Tween-20 Surfact-Amps Detergent Solution is a highly-purified Tween™ 20 detergent stabilized as a 10% solution (w/v) and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring it's stability and eliminating the accumulation of peroxides and degradation products.

Features of Tween-20 Surfact-Amps Detergent Solution:

• Tween 20—a popular nonionic detergent for use in ELISA, Western blotting and other immunoassay PBS or TBS wash buffers
• Accurate—precise 10% detergent solution in ultrapure water
• Easy-to-use—solution is simple to dispense and dilute for use
• Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
• Stable—packaged under inert nitrogen gas in glass ampules or HDPE bottles

Properties of Tween 20:
• Molecular Weight: 1228g
• Detergent Class: Nonionic polyoxyethylene surfactant
• Aggregation Number: Unknown
• Micelle Molecular Weight: Unknown
• Critical Micelle Concentration (CMC): 0.06 mM (0.0074%, w/v)
• Cloud Point: 95°C
• Dialyzable: No

Specifications for Tween 20 Surfact-Amps Detergent Solution:
• Visual: Clear, light yellow liquid, free of particulates.
• Concentration: 10.0±1.0%
• Oxidants: ≤1.0μeq/mL
• Carbonyls: ≤1.0μeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

Related Products
Surfact-Amps™ Detergent Sampler
For Research Use Only. Not for use in diagnostic procedures.
사양
수량6 x 10 mL
시약 유형Detergent Solution
형태Liquid
제품라인Surfact-Amps, Tween
제품 유형Detergent
Unit SizeEach
구성 및 보관
Store in a cool, dry, and well-ventilated area

자주 묻는 질문(FAQ)

What is the Thermo Scientific Tween-20 Surfact-Amps Detergent Solution?

Tween-20 Surfact-Amps Detergent Solution is a highly-purified Tween 20 detergent stabilized as a 10% solution and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring its stability and eliminating the accumulation of peroxides and degradation products.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What quality testing is performed for your Tween 20 Surfact-Amps Detergent Solution?

The detergent is screened for:
- Visualization: clear, light yellow liquid, free of particulates
- Concentration: 10.0 +/- 1.0%
- Oxidant concentration: less than or equal to 1.0 µeq/mL
- Carbonyl concentration: less than or equal to 1.0 µeq/mL
- suspended solids: residue present must not exceed Residue Reference

See the COA for further details.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

인용 및 참조 문헌 (1)

인용 및 참조 문헌
Abstract
Analysis of global gene expression profiles activated by chemoattractant receptors.
Authors:Martinez FO,Locati M
Journal:Methods in molecular biology (Clifton, N.J.)
PubMed ID:16878702
Microarrays are made by immobilizing to a solid support thousands of DNA probes that detect soluble complementary target sequences using the hybridization pairing rules of nucleic acids. Receptor triggering induces a cascade of signaling events that often involves the modulation of gene expression. In the last decade, the development of ... More