Tween™ 20 Surfact-Amps™ Detergent Solution, 6 x 10 mL - FAQs
View additional product information for Tween™ 20 Surfact-Amps™ Detergent Solution - FAQs (28320, 85115, 28321, 85114, 85113)
2 product FAQs found
What is the Thermo Scientific Tween-20 Surfact-Amps Detergent Solution?
Tween-20 Surfact-Amps Detergent Solution is a highly-purified Tween 20 detergent stabilized as a 10% solution and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring its stability and eliminating the accumulation of peroxides and degradation products.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What quality testing is performed for your Tween 20 Surfact-Amps Detergent Solution?
The detergent is screened for:
- Visualization: clear, light yellow liquid, free of particulates
- Concentration: 10.0 +/- 1.0%
- Oxidant concentration: less than or equal to 1.0 µeq/mL
- Carbonyl concentration: less than or equal to 1.0 µeq/mL
- suspended solids: residue present must not exceed Residue Reference
See the COA for further details.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.