Triton™ X-114 Surfact-Amps™ Detergent Solution
Thermo Scientific™

Triton™ X-114 Surfact-Amps™ Detergent Solution

Thermo Scientific Triton X-114 Surfact-Amps Detergent Solution is a stabilized 10% (w/v) solution of highly-purified Triton™ X-114 detergent, supplied inRead more
Have Questions?
Catalog NumberQuantity
283326 x 10 mL
Catalog number 28332
Price (EUR)
436,00
Each
Quantity:
6 x 10 mL
Request bulk or custom format
Price (EUR)
436,00
Each
Thermo Scientific Triton X-114 Surfact-Amps Detergent Solution is a stabilized 10% (w/v) solution of highly-purified Triton™ X-114 detergent, supplied in glass ampules and packaged under nitrogen inert gas, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Features of Triton X-114 Surfact-Amps Detergent Solution:

• Triton X-114—a nonionic detergent for use in various routine protein and molecular biology techniques
• Accurate—precise 10% detergent solution in ultrapure water
• Easy-to-use—solution is simple to dispense and dilute for use
• Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
• Stable— packaged in glass ampules under inert nitrogen gas

Specifications for Triton X-114 Surfact-Amps Detergent Solution:
• Visual: Slightly hazy to cloudy, colorless solution, free of particulates; may form layers.
• Concentration: 10.0±1.0%
• Oxidants: ≤1.0μeq/mL
• Carbonyls: ≤1.0μeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

Properties of Triton X-114:
• Molecular Weight: 537g
• Detergent Class: Nonionic polyoxyethylene surfactant
• Aggregation Number: unknown
• Micelle Molecular Weight: unknown
• Critical Micelle Concentration (CMC): 0.21 mM (0.0113%, w/v)
• Cloud Point: 23°C
• Dialyzable: No

Triton X-114 is most frequently used as a component of cell lysis buffers. In certain conditions, this detergent phase-separates in aqueous solutions, a property that has been used to separate hydrophilic and hydrophobic (membrane) proteins.

For added safety when opening glass ampules, consider using our Ampule Breakers, which are disposable safety devices that protect the fingers when breaking open a glass ampule.

Related Products
Surfact-Amps™ Detergent Sampler
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity6 x 10 mL
Reagent TypeDetergent Solution
FormLiquid
Product LineSurfact-Amps, Triton
Product TypeDetergent
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.

Frequently asked questions (FAQs)

What are the main advantages of using Thermo Scientific Pierce Surfact-Amps detergents?

Thermo Scientific Pierce Surfact-Amps detergents are highly purified, precisely diluted (10%) formulations that are ideal for applications or assays that are sensitive to contaminants that are present in unpurified detergents. We test every batch to insure that our detergents contain less than 1.0 µeq/mL peroxides and carbonyls and package them under nitrogen, to prevent oxidization during storage.

Superior quality - lower measurable contaminant levels than other leading vendors
Accurate -precise 10% detergent solution in ultrapure water
Easy-to-use - solution is simple to dispense and dilute for use
Exceptionally pure - less than 1.0 µeq/mL peroxides and carbonyls

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Are detergents denaturing or non-denaturing with respect to protein structure?

Ionic detergents, or those that carry a charge, are the most likely to be denaturing to proteins. Denaturing detergents can be anionic such as sodium dodecyl sulfate (SDS) or cationic such as ethyl trimethyl ammonium bromide. These detergents totally disrupt membranes and denature proteins by breaking protein-protein interactions through changes in the three-dimensional structure of the proteins. Nondenaturing detergents can be divided into nonionic detergents (i.e., Triton X-100), bile salts (i.e., cholate), and zwitterionic detergents (i.e., CHAPS).



Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What are detergents?

Detergents are amphipathic molecules containing both a nonpolar “tail” having aliphatic or aromatic character, and a polar “head”. The ionic character of the polar head group forms the basis for broad classification of detergents as ionic, nonionic, or zwitterionic.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How does detergent-based cell lysis work?

Detergents are amphipathic molecules, meaning they contain both a nonpolar “tail” having aliphatic or aromatic character and a polar “head”. Like the components of biological membranes, detergents have hydrophobic-associating properties as a result of their nonpolar tail groups. Nevertheless, detergents are themselves water soluble.

Consequently, detergent molecules allow the dispersion (miscibility) of water-insoluble, hydrophobic compounds into aqueous media, including the extraction and solubilization of membrane proteins. Detergent monomers solubilize membrane proteins by partitioning into the membrane bilayer. With increasing amounts of detergents, membranes undergo various stages of solubilization.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What types of detergents are available for cell lysis?

Detergents can be denaturing or non-denaturing with respect to protein structure. Denaturing detergents can be anionic such as sodium dodecyl sulfate (SDS) or cationic such as ethyl trimethyl ammonium bromide. These detergents totally disrupt membranes and denature proteins by breaking proteinprotein interaction. These detergents are considered harsh. Non-denaturing detergents can be divided into nonionic detergents (i.e., Triton X-100), bile salts (i.e., cholate), and zwitterionic detergents (i.e., CHAPS). These detergents do not denature proteins and do not break protein-protein interactions. These detergents are considered mild.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.