NeutrAvidin Protein, HRP - FAQs
NeutrAvidin Protein, HRP - FAQs
View additional product information for NeutrAvidin Protein, HRP - FAQs (31001)
1 product FAQs found
What is the shelf life of NeutrAvidin Protein, HRP (Cat. No. 31001)?
The NeutrAvidin Protein, HRP is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (at 4 degrees C upon receipt). Please see section 8.1 of our Terms & Conditions of Sale for more details.