Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Secondary Antibodies ›
          • Goat anti-Rat IgG Secondary Antibodies

          Invitrogen

          Goat anti-Rat IgG (H+L) Secondary Antibody, HRP

          27 Published Figures
          View all (96) Goat anti-Rat IgG antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support

          Cite Goat anti-Rat IgG (H+L) Secondary Antibody, HRP

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (8)
          • Published Figures (27)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 35

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Rat IgG (H+L) Secondary Antibody (31470) in WB

          Chemiluminescent western blot was performed using Goat anti-Rat IgG (H+L) Secondary Antibody, HRP (Product # 31470). Whole cell extracts of NCCIT (Lane 1, 2, 3, 4), NTERA2 (Lane 5, 6, 7, 8) and HeLa (Lane 9) were electrophoresed usingNuPAGE™ 10% Bis-Tris Protein Gel (Product # NP0301BOX). Resolved proteins were transferred onto anitrocellulose membrane (Product # IB23001) byiBlot® 2 Dry BlottingSystem (Prod... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Rat IgG (H+L) Secondary Antibody in ELISA (ELISA)
          Rat IgG (H+L) Secondary Antibody in ELISA (ELISA)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Western Blot (WB)
          Rat IgG (H+L) Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Product Details

          31470

          Applications
          Tested Dilution

          Western Blot (WB)

          1:5,000-1:200,000

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000

          ELISA (ELISA)

          1:5,000-1:10,000

          Immunoprecipitation (IP)

          1:500-1:5,000
          Product Specifications

          Species Reactivity

          Rat

          Host/Isotype

          Goat / IgG

          Class

          Polyclonal

          Type

          Secondary Antibody

          Conjugate

          HRP HRP HRP
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 555 
          • Alexa Fluor Plus 647 
          • Alexa Fluor Plus 680 
          • AP
          • Biotin 
          • FITC
          • PE 
          • Rhodamine
          • TRITC
          • Unconjugated
          • Request custom conjugation

          Form

          Lyophilized

          Concentration

          0.8 mg/mL

          Amount

          1.6 mg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.6, with 15mg/mL BSA

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_228356

          Target

          IgG

          Antibody Form

          Whole Antibody

          Product Specific Information

          This antibody has been successfully used in Western blot, IHC, ELISA and IP applications.

          Antibody Specificity: This antibody reacts with the heavy chains of rat IgG and with the light chains common to most rat immunoglobulins, based on immunoelectrophoresis. No antibody was detected against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species.

          Restoration and Storage: Store product at 4°C until opened. Restore with 2.0 mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day.

          To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C.

          Country of Origin: USA

          Target Information

          Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          View product citations for antibody 31470 on CiteAb

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0