ABC Peroxidase Standard Staining Kit - FAQs
View additional product information for ABC Peroxidase Standard Staining Kit - FAQs (32020)
1 product FAQs found
Can you provide the shelf life for ABC Peroxidase Standard Staining Kit?
ABC Peroxidase Standard Staining Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.