Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • IFN beta Antibodies

          Invitrogen

          IFN beta Polyclonal Antibody

          1 Published Figure
          1 Reference
          View all (31) IFN beta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IFN beta Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (1)
          IFN beta Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          IFN beta Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IFN beta Antibody (324001) in ELISA

          BMC microbiology 2015 - Fig. 3 Cytokine induction in BMDM infected with C. pneumoniae strains AO3 or AR39. BMDMs were prepared as described in the Methods. The cells were infected with AO3 or AR39 at the indicated MOI, and incubated at 35 degC or 37 degC. Supernatant was harvested at 24 h post infection and assayed for ( a ) IL-1beta, ( b ) IL-6 and ( c ) IFN-beta. Graphs depict the mean +- SEM, and are representative of 3 indepen... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IFN beta Antibody in ELISA (ELISA)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          324001

          Applications
          Tested Dilution
          Publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Neutralization (Neu)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant mouse interferon beta

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum, PBS

          Contains

          no preservative

          Storage conditions

          -80°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_223494

          Product Specific Information

          Specificity: Neutralizes mouse interferon beta.



          Activity: 1.55x10^6 NU/mL. Interferon was titrated with the use of the cytopathic effect inhibition assay using L929 cells with EMCV, where 2.5 unit/mL of interferon was necessary to produce an endpoint of 50%. One neutralization unit is defined as the amount of serum required to neutralize one unit of mouse IFN beta to an endpoint of 50%.



          For retention of full activity, do not store above -70°C.



          Packaged by and distributed for PBL Assay Science.

          Target Information

          Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: IFN b; IFN β; IFNbeta; IFNβ; Interferon; interferon 1da1; Interferon b; RP11-113D19.1

          View more View less

          Gene Aliases: If1da1; Ifb; IFN-beta; IFNB

          View more View less

          UniProt ID: (Mouse) P01575

          View more View less

          Entrez Gene ID: (Mouse) 15977

          View more View less

          Function(s)
          cytokine activity cytokine receptor binding type I interferon receptor binding chloramphenicol O-acetyltransferase activity
          Process(es)
          adaptive immune response macrophage activation involved in immune response T cell activation involved in immune response B cell activation involved in immune response natural killer cell activation involved in immune response negative regulation of immunoglobulin production negative regulation of immune system process regulation of transcription from RNA polymerase II promoter defense response immune response humoral immune response signal transduction JAK-STAT cascade negative regulation of cell proliferation positive regulation of autophagy cytokine-mediated signaling pathway negative regulation of interferon-gamma production cellular response to interferon-beta B cell proliferation defense response to bacterium response to exogenous dsRNA negative regulation of viral genome replication innate immune response leukocyte activation negative regulation of T cell differentiation negative regulation of osteoclast differentiation positive regulation of transcription from RNA polymerase II promoter negative regulation of multicellular organismal process defense response to virus type I interferon signaling pathway negative regulation of cell adhesion molecule production neuron cellular homeostasis cellular response to dsRNA cellular response to dexamethasone stimulus positive regulation of transforming growth factor beta production negative regulation of mononuclear cell migration receptor signaling pathway via STAT cellular response to virus negative regulation of Lewy body formation antiviral innate immune response negative regulation of neuroinflammatory response positive regulation of protein localization to nucleus negative regulation of matrix metallopeptidase secretion negative regulation of blood-brain barrier permeability negative regulation of T-helper 2 cell cytokine production positive regulation of apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0