Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • SRSF3 Antibodies

          Invitrogen

          SRSF3 Monoclonal Antibody (7B4 (7B4A12))

          Advanced Verification
          25 Published Figures
          21 References
          View all (13) SRSF3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SRSF3 Monoclonal Antibody (7B4 (7B4A12))

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (25)
          • Advanced Verification (3)
          SRSF3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SRSF3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 30

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SRSF3 Antibody (33-4200) in WB

          Western blot analysis was performed on nuclear enriched extracts (30 µg lysate) of HeLa (Lane 1), HCT 116 (Lane 2), HT-29 (Lane 3), A549 (Lane 4) and Hep G2 (Lane 5). The blot was probed with Anti-SRP20 Mouse Monoclonal Antibody (Product # 33-4200, 2 µg/mL) and detected by chemiluminescence using Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, HRP conjµgate (Product # A28177, 0.4 µg/mL, 1:2500 dilution). A ~25 kDa band corresponding to SRP20 was observed across the cell lines tested. Known quantity of protein samples were electr... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Immunocytochemistry (ICC/IF)
          SRSF3 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Western Blot (WB)
          SRSF3 Antibody in Immunoprecipitation (IP)
          SRSF3 Antibody
          SRSF3 Antibody
          SRSF3 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SRSF3 Monoclonal Antibody (7B4 (7B4A12))

          Product Details

          33-4200

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          View 15 publications 15 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 2 publications 2 publications

          ELISA (ELISA)

          Assay-dependent
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human, Rat, Xenopus

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          7B4 (7B4A12)

          Immunogen

          A synthetic peptide corresponding to amino acids 84-104 of the human SRp20 protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533119

          Target Information

          SR proteins appear to be recruited from nuclear "speckles", in which they are concentrated, to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: epididymis secretory sperm binding protein; pre-mRNA splicing factor SRp20; pre-mRNA-splicing factor SRP20; RNA binding protein; splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich, 20-kD; unnamed protein product

          View more View less

          Gene Aliases: SFRS3; SRp20

          View more View less

          UniProt ID: (Human) P84103

          View more View less

          Entrez Gene ID: (Human) 6428

          View more View less

          Function(s)
          nucleic acid binding RNA binding mRNA binding protein binding pre-mRNA binding phospholipase binding primary miRNA binding protein-RNA sequence-specific adaptor activity sequence-specific mRNA binding
          Process(es)
          regulation of alternative mRNA splicing, via spliceosome RNA processing mRNA export from nucleus primary miRNA processing regulation of mRNA splicing, via spliceosome
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-xbl4w:80/100.66.75.228:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0