Nalgene™ PPCO Micro Packaging Vial Closures for 4.5 mL Vials: Sterile, Bulk Pack, white - FAQs
View additional product information for Nalgene™ PPCO Micro Packaging Vial Closures for 4.5mL Vials: Sterile, Bulk Pack - FAQs (342826-0110, 342826-0111, 342826-0114)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.