Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Connexin 32 Antibodies

          Invitrogen

          Connexin 32 Monoclonal Antibody (5F9A9)

          40 Published Figures
          35 References
          View all (22) Connexin 32 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Connexin 32 Monoclonal Antibody (5F9A9)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (40)
          Connexin 32 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Connexin 32 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 40

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Connexin 32 Antibody (35-8900) in ICC/IF

          PLoS neglected tropical diseases 2019 - Fig 4 Concentration of intracellular connexin 26, connexin 32, and connexin 43 in human cholangiocytes (H69 cells). After treatment with NDMA and ESP for 72 h, the concentration of intracellular connexin 26, 32, and 43 in H69 cells was measured by laser scanning microscopy (x4,000). A-C: Immunofluorescence confocal microscopy indicated that the intracellular concentrations of Cx26 (green)/DAPI (blue), Cx32 ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Connexin 32 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 32 Antibody in Immunohistochemistry, Immunohistochemistry (Frozen) (IHC, IHC (F))
          Connexin 32 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 32 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 32 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 32 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 32 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 32 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (IHC)
          Connexin 32 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Connexin 32 Antibody in Western Blot (WB)
          Connexin 32 Antibody in Western Blot (WB)
          Connexin 32 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Connexin 32 Antibody in Western Blot (WB)
          Connexin 32 Antibody in Western Blot (WB)
          Connexin 32 Antibody in Western Blot (WB)
          Connexin 32 Antibody in Western Blot (WB)
          Connexin 32 Antibody in Western Blot (WB)
          Connexin 32 Antibody in Western Blot (WB)
          Connexin 32 Antibody in Western Blot (WB)
          Connexin 32 Antibody in Western Blot (WB)
          Connexin 32 Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Connexin 32 Monoclonal Antibody (5F9A9)

          Product Details

          35-8900

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 8 publications 8 publications

          Immunohistochemistry (IHC)

          -
          View 12 publications 12 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 4 publications 4 publications

          Immunoprecipitation (IP)

          -
          View 3 publications 3 publications

          Miscellaneous PubMed (Misc)

          -
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          5F9A9

          Immunogen

          Synthetic peptide derived from the C-terminus of mouse Connexin 32.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533228

          Target Information

          This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: beta-a gap junction protein; connexin 32; connexin g; connexin-32; connexin-43; GAP junction 28 kDa liver protein; gap junction membrane channel protein beta 1; unnamed protein product

          View more View less

          Gene Aliases: Cnx32; connexin-32; Cx32; Cxng

          View more View less

          UniProt ID: (Mouse) P28230, (Rat) P08033

          View more View less

          Entrez Gene ID: (Mouse) 14618, (Rat) 29584

          View more View less

          Function(s)
          gap junction channel activity identical protein binding
          Process(es)
          cell communication cell-cell signaling purine ribonucleotide transport transmembrane transport epididymis development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch