Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack, 0.5mL - FAQs
View additional product information for Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack - FAQs (362805-0020, 362805-0005)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.