Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack
Thermo Scientific™

Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack

Protect light-sensitive reagents with these PPCO amber micro packaging vials featuring conical bottoms that allow recovery of entire contents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
362805-00050.5 mL
362805-00202 mL
Catalog number 362805-0005
Price (USD)
182.57
Online exclusive
202.86
Save 20.29
Case of 1000
Volume (Metric):
0.5 mL
Price (USD)
182.57
Online exclusive
202.86
Save 20.29
Case of 1000
Protect light sensitive reagents with Thermo Scientific™ Nalgene™ PPCO Amber Micro Packaging Vials. These noncytotoxic and nonpyrogenic vials are molded from high purity, low-metal content polypropylene copolymer resin. They are skirted and feature a conical bottom, allowing recovery of entire contents.
Features:
  • Vials and closures (listed separately) pressure-tested together at 7.5 psig
  • USP Class VI compliant; nonpyrogenic
  • Can be centrifuged at 13,000 x g
  • Colored closures packaged separately
  • Nonsterile
  • Leakproof† when used with Nalgene closures

Ordering Information:

Vials are bulk packed without closures, 500 per inner bag, 1,000 per case. Order nonsterile closures separately (Cat. Nos. 362821, 362825).

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Certifications/ComplianceISO 13485:2016
Closure MaterialPPCO
Closure Size11 mm
DescriptionMicro Packaging Vials
Diameter (Metric) Inner8 mm
Height (Metric)49 mm
No. per Case1000
No. per Pack500
Product TypeVial
Volume (Metric) Total0.5 mL
Closure TypeScrew Cap (Not Included)
ColorAmber
Diameter (Metric) Outer13 mm
MaterialPPCO
Quantity500 Bag, 1000 Cs.
SterilityNon-sterile
Volume (Metric)0.5 mL
Unit SizeCase of 1000

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.