Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.
Search
View additional product information for Nalgene™ PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile - FAQs (362820-0117, 362820-0116, 362820-0119, 362820-0118, 362820-0113, 362820-0112, 362820-0115, 362820-0114, 362820-0111, 362820-0110, 362820-1110)
1 product FAQs found