Search
View additional product information for Nunc™ Universal and Transport Containers with Caps - FAQs (363282, 364882, 364246, 364238)
1 product FAQs found
Yes, the Nunc Transport System consists of two containers. The inner vial or Nunc Universal Container is designed to fit inside of the outer vial or Nunc Transport Container. This dual system is designed to provide safe and secure handling for transport of biological specimens.