Search
View additional product information for Phospho-ERK1/ERK2 (Thr202, Tyr204) Polyclonal Antibody - FAQs (368800)
1 product FAQs found
Yes, we have many antibodies against ERK 1 and 2 (e.g. 44-654G, 136200) and their phosphorylated forms. Examples of the latter include Cat. Nos. 44680G, 700012, 368800, and PA14607, among many others .
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.