Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • EphB2 Antibodies

          Invitrogen

          EphB2 Monoclonal Antibody (1A6C9)

          Advanced Verification
          11 Published Figures
          16 References
          View all (26) EphB2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite EphB2 Monoclonal Antibody (1A6C9)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (11)
          • Advanced Verification (4)
          EphB2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          EphB2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 17

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EphB2 Antibody (37-1700) in WB

          Western blot was performed using Anti-EphB2 Monoclonal Antibody (1A6C9) (Product # 37-1700) and 117, 110kDa bands corresponding to EphB2 was increased upon VEGF treatment in HUVEC. Also, the expression was highest in COLO 205 in comparison to SW480 and HT-29 as reported. Membrane enriched cell extracts (30 ug lysate) of HUVEC (Lane 1), HUVEC treated with VEGF (20ng/ml, 10 min) (Lane 2), COLO 205 (Lane 3), SW480 (Lane 4) and HT-29 (Lane 5) were electrophoresed using Novex® NuPAGE® 4-12 % Bis-Tris gel (Product # NP0322BOX). Resolved proteins... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EphB2 Antibody in Western Blot (WB)
          EphB2 Antibody in Western Blot (WB)
          EphB2 Antibody in Western Blot, Immunocytochemistry, Immunoprecipitation (WB, ICC/IF, IP)
          EphB2 Antibody in Western Blot (WB)
          EphB2 Antibody in Western Blot (WB)
          EphB2 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          EphB2 Antibody in Western Blot (WB)
          EphB2 Antibody in Western Blot (WB)
          EphB2 Antibody in Western Blot (WB)
          EphB2 Antibody in Western Blot (WB)
          EphB2 Antibody in Western Blot (WB)
          EphB2 Antibody in Western Blot (WB)
          EphB2 Antibody in Western Blot (WB)
          EphB2 Antibody
          EphB2 Antibody
          EphB2 Antibody
          EphB2 Antibody
          EphB2 Monoclonal Antibody (1A6C9)

          Product Details

          37-1700

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          View 9 publications 9 publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          View 3 publications 3 publications

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Chicken, Human

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1A6C9

          Immunogen

          A synthetic peptide derived from the C-terminal region of the recombinant EphB2 protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533302

          Target Information

          EPHB2 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cEK5; chicken embryo kinase 5; Developmentally-regulated Eph-related tyrosine kinase; EFL6; EFNB3; EK5; ELK; ELK-related tyrosine kinase; embryo kinase 5 protein CEK5; EPH B2; EPH tyrosine kinase 3; EPH-like kinase 5; EPHB3; Ephrin B1; Ephrin B2; Ephrin B3; Ephrin B4; Ephrin type-B receptor 2; EphrinB1; EphrinB2; EphrinB3; EphrinB4; EPLG8; ETK2; HEK-6; HEK2; LERK8; MGC87492; NET; protein-tyrosine kinase HEK5; Renal carcinoma antigen NY-REN-47; TYRO6; Tyrosine-protein kinase receptor EPH-3; Tyrosine-protein kinase TYRO5; unnamed protein product

          View more View less

          Gene Aliases: BDPLT22; CAPB; CEK5; DRT; EK5; EPHB2; EPHT3; EPTH3; ERK; HEK5; PCBC; TYRO5

          View more View less

          UniProt ID: (Human) P29323, (Chicken) P28693

          View more View less

          Entrez Gene ID: (Human) 2048, (Chicken) 396513

          View more View less

          Function(s)
          nucleotide binding beta-amyloid binding protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity ephrin receptor activity transmembrane-ephrin receptor activity receptor binding protein binding ATP binding axon guidance receptor activity kinase activity transferase activity signaling receptor activity identical protein binding macromolecular complex binding transmembrane signal receptor
          Process(es)
          cell morphogenesis angiogenesis urogenital system development positive regulation of immunoglobulin production negative regulation of cell adhesion transmembrane receptor protein tyrosine kinase signaling pathway nervous system development axonogenesis axon guidance axonal fasciculation learning or memory learning organ morphogenesis positive regulation of gene expression phosphorylation peptidyl-tyrosine phosphorylation optic nerve morphogenesis hindbrain tangential cell migration central nervous system projection neuron axonogenesis corpus callosum development regulation of blood coagulation positive regulation of cell migration positive regulation of B cell proliferation retinal ganglion cell axon guidance positive regulation of synaptic plasticity positive regulation of tumor necrosis factor production B cell activation inner ear morphogenesis regulation of JAK-STAT cascade negative regulation of Ras protein signal transduction ephrin receptor signaling pathway regulation of neuronal synaptic plasticity positive regulation of long-term neuronal synaptic plasticity camera-type eye morphogenesis regulation of axonogenesis negative regulation of axonogenesis regulation of body fluid levels positive regulation of multicellular organismal process regulation of filopodium assembly regulation of synapse assembly positive regulation of synapse assembly palate development dendritic spine development dendritic spine morphogenesis positive regulation of dendritic spine morphogenesis negative regulation of ERK1 and ERK2 cascade cellular response to lipopolysaccharide commissural neuron axon guidance postsynaptic membrane assembly neuron projection retraction vesicle-mediated intercellular transport tight junction assembly negative regulation of cytokine production involved in inflammatory response positive regulation of long-term synaptic potentiation negative regulation of glutamate receptor signaling pathway positive regulation of glutamate receptor signaling pathway positive regulation of protein localization to plasma membrane cellular response to beta-amyloid neuron projection maintenance positive regulation of protein localization to cell surface regulation of T-helper 17 type immune response regulation of autophagosome assembly regulation of behavioral fear response multicellular organism development spinothalamic tract morphogenesis positive regulation of kinase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline