Matrix™ 1.1 mL Screw-Top Tubes - FAQs
Matrix™ 1.1 mL Screw-Top Tubes - FAQs
View additional product information for Matrix™ ScrewTop Tri-coded Tubes - FAQs (3741NS-WP1D-BR, 3743-WP1D, 3744-WP1D-BR, 3740-WP1D, 3745-WP1D-BR, 3742-WP1D, 3745-WP1D, 3744-WP1D, 3742NS-WP1D, 3742-WP1D-BR, 3745NS-WP1D-BR, 3744NS-WP1D-BR, 3741-WP1D-BR, 3741-WP1D)
1 product FAQs found
What is the Thermo Fisher alternative to Corning® 2 mL Internal Threaded Polypropylene Cryogenic Vial with 1D and 2D Bar codes, Product Number 8670?
The following alternatives for the Corning Cryogenic Vial, Product Number 8670:
- Nunc 2.0 mL Internally-Threaded Universal Tubes (Cat. No. 374510, 374510-BR)
- Nunc 2.0 mL Internally-Threaded Universal Tubes (Cat. No. 374512). Please note that this product is FDA registered and CE-marked.
- Nunc 1.8mL Externally-Threaded Universal Tubes and Racks (Cat. No. 374502, 374502NOV
- Matrix ScrewTop Tri-coded Tubes (Cat. No. 3740-WP1D)