Matrix™ 2D Barcoded Low Volume ScrewTop Storage Tubes, Storage tube w/ 2D code in barcoded rack - FAQs
View additional product information for Matrix™ 2D Barcoded Low Volume ScrewTop Storage Tubes - FAQs (3748, 3748-BR, 3747)
Error:This product does not have any FAQs available.