Phtalate de dicyclohexyle, 99 %, Thermo Scientific Chemicals
Phtalate de dicyclohexyle, 99 %, Thermo Scientific Chemicals
Phtalate de dicyclohexyle, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phtalate de dicyclohexyle, 99 %, Thermo Scientific Chemicals

CAS: 84-61-7 | C20H26O4 | 330.42 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
5 g
250 g
1 kg
2.5 kg
Conditionnement:
Flacon en verre
Référence 375050010
Prix (EUR)
169,00
Each
Quantité:
1 kg
Conditionnement:
Flacon en verre
Grand volume ou format personnalisé
Prix (EUR)
169,00
Each
Identifiants chimiques
CAS84-61-7
IUPAC Name1,2-dicyclohexyl benzene-1,2-dicarboxylate
Molecular FormulaC20H26O4
InChI KeyVOWAEIGWURALJQ-UHFFFAOYSA-N
SMILESO=C(OC1CCCCC1)C1=CC=CC=C1C(=O)OC1CCCCC1
Voir plus
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White
Melting point63°C to 67°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only