Search
View additional product information for 250mL Conical Centrifuge Tube - FAQs (376814)
1 product FAQs found
Yes, the same cushion adapter (Cat. No. 377585) can be used for both bottles because they are predominantly the same shape (including the lower conical section).