Nunc™ Centrifuge Bottle Adapters and Cushions - FAQs
Nunc™ Centrifuge Bottle Adapters and Cushions - FAQs
View additional product information for Nunc™ Centrifuge Bottle Adapters and Cushions - FAQs (377585)
1 product FAQs found
Do the Nunc 200 mL (Cat. No. 376813) and 250 mL (Cat. No. 376814) sterile conical centrifuge bottles use the same adapter?
Yes, the same cushion adapter (Cat. No. 377585) can be used for both bottles because they are predominantly the same shape (including the lower conical section).