Lead(II) sulfate, 99%
Lead(II) sulfate, 99%
Lead(II) sulfate, 99%
Lead(II) sulfate, 99%
Thermo Scientific Chemicals

Lead(II) sulfate, 99%

CAS: 7446-14-2 | O4PbS | 303.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 387271000
Price (EUR)
30,65
Online exclusive
34,30
Save 3,65 (11%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
30,65
Online exclusive
34,30
Save 3,65 (11%)
Each
Chemical Identifiers
CAS7446-14-2
IUPAC Nameλ²-lead(2+) sulfate
Molecular FormulaO4PbS
InChI KeyKEQXNNJHMWSZHK-UHFFFAOYSA-L
SMILES[Pb++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Elemental Analysis>=98.5 % (AES)
Appearance (Form)Crystalline powder
IdentificationConfirmed

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only