ANGPTL4 Polyclonal Antibody - FAQs
ANGPTL4 Polyclonal Antibody - FAQs
View additional product information for ANGPTL4 Polyclonal Antibody - FAQs (409800)
1 product FAQs found
Can you please let me know whether the immunogen sequence of the ANGPTL4 Polyclonal Antibody (Cat. No. 40-9800) is before or after the 168 amino acid of the protein?
The immunogen used was from amino acids 259-274 of the protein sequence in Uniprot ID Q9BY76.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.