Itaconic anhydride, 98%
Itaconic anhydride, 98%
Itaconic anhydride, 98%
Thermo Scientific Chemicals

Itaconic anhydride, 98%

CAS: 2170-03-8 | C5H4O3 | 112.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 412940250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2170-03-8
IUPAC Name3-methylideneoxolane-2,5-dione
Molecular FormulaC5H4O3
InChI KeyOFNISBHGPNMTMS-UHFFFAOYSA-N
SMILESC=C1CC(=O)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Infrared spectrumConforms
Assay>=97.5 %
Appearance (Form)Crystalline powder
Melting point66°C to 70°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only