Thioacetamide, 99+%, ACS reagent
Thioacetamide, 99+%, ACS reagent
Thioacetamide, 99+%, ACS reagent
Thermo Scientific Chemicals

Thioacetamide, 99+%, ACS reagent

CAS: 62-55-5 | C2H5NS | 75.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 424535000
Price (TWD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS62-55-5
IUPAC Nameethanethioamide
Molecular FormulaC2H5NS
InChI KeyYUKQRDCYNOVPGJ-UHFFFAOYSA-N
SMILESCC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point111°C to 114°C
Titration Argentometric>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only