4-tert-Butylbenzaldehyde, 95%
4-tert-Butylbenzaldehyde, 95%
4-tert-Butylbenzaldehyde, 95%
Thermo Scientific Chemicals

4-tert-Butylbenzaldehyde, 95%

CAS: 939-97-9 | C11H14O | 162.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 426522500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS939-97-9
IUPAC Name4-tert-butylbenzaldehyde
Molecular FormulaC11H14O
InChI KeyOTXINXDGSUFPNU-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5270 to 1.5290 (20°C, 589 nm)
GC>=94 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only