Are the BigDye Terminator 5X buffer v1.1 & v3.1 and BigDye Terminator 5X buffer v1.0 & v3.0 interchangeable?
The composition of the BigDye Terminator 5X buffer v1.1 & v3.1 and BigDye Terminator 5X buffer v1.0 & v3.0 chemistries is different and therefore should only be used with the matching chemistry. Use of the incorrect BigDye Terminator 5X buffer can negatively impact sequencing results.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.