Product Image
Applied Biosystems™

StockMarks™ for Cattle Genotyping Kit, bovine, with or without protocol

This PCR-based system is used to genotype bovines and validate their parentage quickly and accurately. The StockMarks™ for Cattle BovineRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
4307480Kit only
4330663Kit with protocol
Catalog number 4307480
Price (ZAR)
-
Includes:
Kit only
This PCR-based system is used to genotype bovines and validate their parentage quickly and accurately. The StockMarks™ for Cattle Bovine Parenting Typing Kit is part of a complete solution for bovine parentage typing and genotyping employing ISAG-recommended microsatellite markers. Protocol is included.

• PCR process provides greater accuracy than serological methods.
• ISAG-recommended, standardized loci ensure accurate testing.
• Proprietary primer purification process ensures optimal data quality.

Accurate with All Sample Types
All StockMarks™ kits amplify microsatellite markers by means of PCR and fluorescent dye-labeled primers. PCR more accurate than serological typing and consumes less sample, is easier to standardize and automate, and is successful with all types of samples, including blood, semen, and hair. Additionally, when fluorescent dye is used to label PCR primers, multiple amplifications can be performed in a single reaction.

Microsatellite-Based Kits Provide Accurate Parentage Testing
The StockMarks™ for Cattle Bovine Parenting Typing Kit both confirms or excludes parentage, using the highly accurate and informative polymorphic short tandem repeats (STRs), also known as microsatellites, which are evenly distributed across the genome.

Used With:
0.5mL GeneAmp™ PCR System 9700, PrepMan™ Ultra Sample Preparation Reagent with Protocol

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodCapillary Electrophoresis
For Use With (Application)Sequencing
For Use With (Equipment)310 Genetic Analyzer, 3100 Genetic Analyzer, 3100-Avant Genetic Analyzer, GeneAmp 9600, GeneAmp 9700
Hot StartBuilt-In Hot Start
IncludesKit only
Label or DyeFAM, JOE, NED
PCR MethodMultiplex PCR
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineStockMarks
Product TypeGenotyping Kit
Quantity1 Kit
Sample TypeDNA
Shipping ConditionDry Ice
SpeciesCattle⁄Bovine
TargetBovine Genotyping and Parentage Testing
Target LociETH3, TGLA122, ETH225, INRA23, TGLA126, BM1824
TechniqueSTR (Short Tandem Repeat) Analysis
FormatKit
Unit SizeEach
Contents & Storage
This kit includes the dye-labeled bovine primer mix, dNTP solution, required reaction buffer, bovine control DNA, and AmpliTaq Gold™ DNA polymerase. The kit contains enough reagents for 100 assays.