MicroAmp™ Optical 96-Well Reaction Plate, 500 Plates - FAQs
MicroAmp™ Optical 96-Well Reaction Plate, 500 Plates - FAQs
View additional product information for MicroAmp™ Optical 96-Well Reaction Plate - FAQs (N8010560, 4316813)
5 product FAQs found
Are the MicroAmp Optical 96-Well Reaction Plates and MicroAmp Optical 384 -Well Reaction Plates autoclavable?
Our microplates are certified to be DNA, RNase and PCR inhibitor free as long as they are kept in the original packaging bag. They are not meant to be autoclaved.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Are the MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737, 4326659) and the one without Barcode (Cat. No. N8010560, 4316813) the same product?
Yes, both the plates are the same. The only difference between them is the barcode.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Can I place a standard MicroAmp 96-Well Optical Reaction Plate onto a 7500 Fast Real-Time PCR System?
No. The plate does not fit and will result in the crushing of the wells and possible contamination of the block due to leaking of the well contents.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Is the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4316813) compatible with the SeqStudio Genetic Analyzer?
Yes, the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4316813) is compatible with the SeqStudo Genetic Analyzer.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Is the 96 well plate in SequalPrep Normalization Plate Kit (Cat. No. A1051001) the same as the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4316813)?
No, they are not the same. The MicroAmp Optical 96-Well Reaction Plate is an empty optical plate that is used for sample processing (i.e. amplification using the thermal cycler or sample loading onto a capillary instrument). The MicroAmp plate cannot be used for performing normalization as it is not coated using the ChargeSwitch technology.