MicroSEQ™ Full Gene 16S rDNA PCR Kit - FAQs
MicroSEQ™ Full Gene 16S rDNA PCR Kit - FAQs
View additional product information for MicroSEQ™ Full Gene 16S rDNA PCR Kit - FAQs (4349155)
22 product FAQs found
What primers are supplied with the MicroSEQ Full Gene 16S rDNA PCR Kit?
The MicroSEQ Full Gene 16S rDNA PCR Kit includes universal domain-level primers that are designed to amplify the bacterial 16S rRNA gene.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
Where can I get a copy of the free trial version of the MicrobeBridge Software?
A 60-day-free trial of the MicrobeBridge Software can be downloaded here: www.thermofisher.com/order/catalog/product/A29238
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Who do I contact for support for MicrobeBridge Software and for MicrobeNet database issues?
For the MicrobeBridge Software, please contact your local Thermo Fisher Scientific Technical Support (https://www.thermofisher.com/us/en/home/technical-resources/contact-us.html) or email abtechsupport@thermofisher.com.
For MicrobeNet, please contact the CDC directly at microbenet@cdc.gov. This account is actively monitored by the CDC MicrobeNet staff.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the minimum system requirements for installing the MicrobeBridge Software?
The MicrobeBridge Software has the following recommended hardware requirements: Minimum 1.8 GHz processor, 2 GB RAM, and 20 GB of free hard disk space. MicrobeBridge Software has been tested on Windows 7 32-bit SP1, Windows 7 64-bit SP1, and Windows 10 IoT.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Will I lose any data if I upgrade from the MicrobeBridge Software free trial version to the full version?
No data is lost with the MicrobeBridge Software installation of the full version on top of the free trial version.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which input file formats can I use with the MicrobeBridge Software?
Only .ab1 files can be used with the MicrobeBridge Software.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which instruments does the MicrobeBridge Software support?
The MicrobeBridge Software fully supports data outputs from Applied Biosystems 3500, 3500xL, 3130, 3130xl, and SeqStudio genetic analyzer systems.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Do I need a MicrobeNet account to use MicrobeNet?
To use MicrobeNet, you will need to register for a MicrobeNet account. You can register at https://microbenet.cdc.gov/
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Is there a list of organisms that can be identified with MicrobeBridge Software?
MicrobeBridge Software does not do bacterial identification. It provides a connection to MicrobeNet which does the bacterial identification.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Can I edit the MicrobeNet software search parameters in MicrobeBridge Software?
No. MicrobeBridge Software does not provide any search parameters which are used by MicrobeNet software.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Can I manually override the changes that the MicrobeBridge Software makes to the contig sequence?
Yes, you can edit the contig sequence generated by the MicrobeBridge software.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Can I export the final contig sequence generated by the MicrobeBridge Software to search in other databases?
Yes, the final contig sequence generated by MicrobeBridge Software can be exported for use in other database searches.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Does MicrobeBridge Software do basecalling?
Yes, MicrobeBridge Software uses Applied Biosystems KB Basecaller v1.4.2.4, by default.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Do I need to sequence with a specific chemistry to use MicrobeBridge Software?
As long as the 16S full gene is covered, a specific chemistry is not needed when using the MicrobeBridge Software.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Do you have recommended protocols, reagents, and primers, to generate MicrobeBridge Software samples?
CDC MicrobeNet has recommended primers which have been configured into the MicrobeBridge Software. We recommend using MicroSEQ 16S Full Gene kit for a complete package solution.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Can sequences generated using the MicroSEQ PCR and sequencing kits be analyzed using MicrobeBridge Software?
Data generated from the MicroSEQ 16S Full Gene Kit package can be analyzed using MicrobeBridge Software.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the difference between MicrobeBridge Software and MicroSEQ ID software?
Through a partnership with the Special Bacteriology Reference Laboratory at the CDC, Thermo Fisher Scientific developed the MicrobeBridge Software to enable public health, microbiology, and any other laboratories worldwide to directly access the CDC's MicrobeNet virtual reference database using 16S rRNA sequencing technology.
MicrobeBridge Software provides a streamlined workflow for analyzing Sanger sequencing data (.ab1 data files) generated on CE Sanger sequencing instruments or genetic analyzers. MicrobeBridge Software performs contig assembly, contig editing, primer trimming, data quality check, contig sequence copying, and one-click connection to MicrobeNet for bacterial identification through 16S rRNA BLAST searches.
In contrast, MicroSEQ ID software is part of a complete system designed for pharmaceutical analytics workflows. The system includes secondary analysis software, validated reference libraries for bacterial and fungal identification, links to query public domain databases, creation of custom reference databases, and SAE features that enables 21 CFR Part 11 compliance.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Is MicrobeBridge Software a replacement for the MicroSEQ ID software?
MicrobeBridge Software is not a replacement for the MicroSEQ ID software. MicrobeBridge Software is specifically designed for public health, microbiology, and any other laboratories utilizing the MicrobeNet database. Applied Biosystems MicroSEQ ID Microbial Identification software is designed for use in pharmaceutical analytics laboratories who typically do not use the MicrobeNet database but require other functionalities. However MicroSEQ ID users can install and use MicrobeBridge Software if they wish to do MicrobeNet searches.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Who can benefit from the MicrobeBridge Software?
Public health, microbiology, and any other laboratories conducting disease surveillance or bacterial identification with MicrobeNet can benefit from using the MicrobeBridge Software.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the benefits of using the MicrobeBridge Software?
Some of the benefits of using the MicrobeBridge Software Faster are faster turnaround time and lower cost:
Using MicrobeBridge Software, MicrobeNet users are able to efficiently assemble and QC their sequencing data using the CDC recommended primer set for 16S rRNA gene sequencing analysis. Users are able to get to their results quickly and accurately, thereby saving money and time spent assembling, interrogating, and identifying bacteria or pathogens in their data.
Users can connect to MicrobeNet via a single click in the software or export the consensus contig and this in turn improves the bacterial identification workflows in public health and microbiology laboratories using MicrobeNet.
Users are able to connect directly into the CDC MicrobeNet virtual reference database for identifying rare and complex pathogens faster and more accurately. Although any laboratory with an internet connection can access the MicrobeNet database worldwide, MicrobeBridge Software streamlines the CE data and connects to MicrobeNet directly.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is MicrobeBridge Software?
The Applied Biosystems MicrobeBridge Software is a desktop application which connects .ab1 data files generated on Applied Biosystems 3500 and 3130 Genetic Analyzers and 3730 DNA Analyzers with the Centers for Disease Control and Prevention (CDC) MicrobeNet virtual reference database via internet connection for bacterial identification using 16S rRNA gene sequencing analysis.
MicrobeNet is a web-based reference database for the identification of microbial pathogens through 16S rRNA gene sequence BLAST searches. MicrobeNet contains a database of genetic and phenotypic properties for bacterial identification actively curated by the CDC.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.