Can one use the reagents from High Capacity RT kit to make cDNA for TaqMan MicroRNA Assay?
Yes. You can use the reagents from the High Capacity cDNA Kit PN 4368814 (without RNAse Inhibitor) or 4374966 (with RNAse Inhibitor). Make sure to use the reverse transcription primer provided from the TaqMan miRNA assay for the reverse transcription.