Find Real-Time and Digital PCR technical tips, troubleshooting help, and resources within our new Real-Time PCR and Digital PCR Applications Support Center.
Upgrade to Maxima™ H Minus First Strand cDNA Synthesis Kit for better performance. List price reduced by 40%.
High-Capacity cDNA Reverse Transcription Kit
Applied Biosystems™

High-Capacity cDNA Reverse Transcription Kit

The High Capacity cDNA Reverse Transcription Kit, available with or without RNase Inhibitor, contains all components necessary for the quantitative conversion of up to 2 μg of total RNA into single-stranded cDNA in a single 20 μL reaction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesNo. of Reactions
4368814Kit only200 Reactions
4374966Kit with Rnase Inhibitor200 Reactions
4368813Kit only1,000 Reactions
4374967Kit with Rnase Inhibitor1,000 Reactions
Catalog number 4368814
Price (USD)
546.00
Each
Includes:
Kit only
No. of Reactions:
200 Reactions
Price (USD)
546.00
Each

The High Capacity cDNA Reverse Transcription Kit, available with or without RNase Inhibitor, contains all components necessary for the quantitative conversion of up to 2 μg of total RNA into single-stranded cDNA in a single 20 μL reaction. Both types of kit deliver extremely high-quality, single-stranded cDNA from 0.02–2 μg total RNA. Reactions can be scaled up to 100 μL to generate 10 μg of cDNA from a single reaction.

The High Capacity cDNA Reverse Transcription Kit with RNase Inhibitor includes the recombinant enzyme used to prevent degradation of RNA templates and lacks DNA endonuclease activity for better product yield.

In addition to their use in RT-PCR, this kit may be used in traditional PCR, microarrays, and for the generation of cDNA archives.

Features of the High Capacity cDNA Reverse Transcription Kit include:

  • Linear target amplification for real-time PCR
  • Higher yields and precision than other cDNA synthesis kits at a fraction of the cost
  • 10-fold greater dynamic range than other kits

Extensively tested with a variety of templates

Quantitative first-strand synthesis of all RNA species is achieved with the use of the random primers. This kit has been extensively tested with a variety of RNA templates, including GC- and AU-rich targets and RNAs expressed at low levels. cRNA can also be efficiently generated from in vitro transcription of cDNA.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatKit
IncludesKit only
No. of Reactions200 Reactions
Optimal Reaction Temperature37°C
Quantity1 Ea.
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Reverse TranscriptaseMultiScribe
Ribonuclease H ActivityYes
Shipping ConditionDry Ice
Size (Final Product)7 kb or Less
Starting MaterialRNA
TechniqueReverse Transcription
Concentration50 U/μL
For Use With (Application)RT-PCR
Reaction Speed120 min.
Unit SizeEach
Contents & Storage

• 10X RT Buffer, 1 mL
• 10X RT Random Primers, 1 mL
• 25X dNTP Mix, 0.2 mL (100 mM)
• MultiScribe Reverse Transcriptase, 2 x 0.1 mL (50 U/μL)

Store at –15 to –25°C.

Frequently asked questions (FAQs)

Can one use the reagents from High Capacity RT kit to make cDNA for TaqMan MicroRNA Assay?

Yes. You can use the reagents from the High Capacity cDNA Kit PN 4368814 (without RNAse Inhibitor) or 4374966 (with RNAse Inhibitor). Make sure to use the reverse transcription primer provided from the TaqMan miRNA assay for the reverse transcription.

Which kit should I use for the reverse transcription step for TaqMan Pri-miRNA assays (Cat. No. 4427012)?

We recommend the following kits:

  • High-Capacity cDNA Reverse Transcription Kit (Cat. No. 4374966).
  • High-Capacity RNA-to-cDNA Kit (Cat. No. 4387406).


Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

Is the 25X 100 mM dNTP Mix included in the High Capacity cDNA Reverse Transcription Kit (Cat. No. 4368813, 4368814) 100 mM each or 100 mM combined?

It contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP).

Citations & References (1122)

Citations & References
Abstract
Hyaluronan-CD44-ERK1/2 regulate human aortic smooth muscle cell motility during aging.
Authors:Vigetti D,Viola M,Karousou E,Rizzi M,Moretto P,Genasetti A,Clerici M,Hascall VC,De Luca G,Passi A
Journal:The Journal of biological chemistry
PubMed ID:18077444
Increased hepatobiliary clearance of unconjugated thyroxine determines DMP 904-induced alterations in thyroid hormone homeostasis in rats.
Authors:Wong H,Lehman-McKeeman LD,Grubb MF,Grossman SJ,Bhaskaran VM,Solon EG,Shen HS,Gerson RJ,Car BD,Zhao B,Gemzik B
Journal:Toxicological sciences : an official journal of the Society of Toxicology
PubMed ID:15673846
4-(3-pentylamino)-2,7-dimethyl-8-(2-methyl-4-methoxyphenyl)-pyrazolo-[1,5-a]-pyri midine (DMP 904) is a potent and selective antagonist of corticotropin releasing factor receptor-1 (CRF1 receptor) with an efficacious anxiolytic profile in preclinical animal models. In subchronic toxicity studies in Sprague-Dawley rats, DMP 904 produced thyroid follicular cell hypertrophy and hyperplasia, and a low incidence of follicular cell ... More
Somatic cell plasticity and Niemann-Pick type C2 protein: fibroblast activation.
Authors:Csepeggi C, Jiang M, Kojima F, Crofford LJ, Frolov A
Journal:J Biol Chem
PubMed ID:21084287
A growing body of evidence points toward activated fibroblasts, also known as myofibroblasts, as one of the leading mediators in several major human pathologies including proliferative fibrotic disorders, invasive tumor growth, rheumatoid arthritis, and atherosclerosis. Niemann-Pick Type C2 (NPC2) protein has been recently identified as a product of the second ... More
TGF-beta regulates miR-206 and miR-29 to control myogenic differentiation through regulation of HDAC4.
Authors:Winbanks CE, Wang B, Beyer C, Koh P, White L, Kantharidis P, Gregorevic P
Journal:J Biol Chem
PubMed ID:21324893
MicroRNAs (miRs) are emerging as prominent players in the regulation of many biological processes, including myogenic commitment and skeletal muscle formation. Members of the TGF-beta family can influence the proliferation and myogenic differentiation of cells, although it is presently not clear what role miRNAs play in the TGF-beta-mediated control of ... More
Disordered macrophage cytokine secretion underlies impaired acute inflammation and bacterial clearance in Crohn's disease.
Authors:Smith AM, Rahman FZ, Hayee B, Graham SJ, Marks DJ, Sewell GW, Palmer CD, Wilde J, Foxwell BM, Gloger IS, Sweeting T, Marsh M, Walker AP, Bloom SL, Segal AW
Journal:J Exp Med
PubMed ID:19652016
The cause of Crohn's disease (CD) remains poorly understood. Counterintuitively, these patients possess an impaired acute inflammatory response, which could result in delayed clearance of bacteria penetrating the lining of the bowel and predispose to granuloma formation and chronicity. We tested this hypothesis in human subjects by monitoring responses to ... More