Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • FLK1 (VEGF Receptor 2) Antibodies

          Invitrogen

          Phospho-VEGF Receptor 2 (Tyr1054) Polyclonal Antibody

          5 References
          View all (88) FLK1 (VEGF Receptor 2) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-VEGF Receptor 2 (Tyr1054) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Phospho-VEGF Receptor 2 (Tyr1054) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-VEGF Receptor 2 (Tyr1054) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-VEGF Receptor 2 (Tyr1054) Antibody (44-1046) in WB

          Peptide Competition and Stimulation: Extracts prepared from PAE cells transfected with a chimeric receptor and stimulated with CSF (1-4) or from NIH3T3 cells transfected with human VEGFR2 left unstimulated (5) or stimulated with VEGF (6) were resolved by {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-VEGF Receptor 2 (Tyr1054) Antibody in Western Blot (WB)
          Phospho-VEGF Receptor 2 (Tyr1054) Polyclonal Antibody

          Product Details

          44-1046

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic phosphopeptide from human VEGFR2 containing tyrosine 1054. The sequence is conserved in mouse and rat.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          Dulbecco's PBS, pH 7.3, with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533556

          Target Information

          KDR (FLK1, VEGFR2) is a type III receptor tyrosine kinase. It binds vascular endothelial growth factor with high affinity in vitro, and is expressed early in development by endothelial cell precursors. FLK1 (VEGF Receptor 2) encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of FLK1 (VEGF Receptor 2) are implicated in infantile capillary hemangiomas.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD309; CD309 antigen; Fetal liver kinase 1; fetal liver kinase-1; FLK-1; KDR; Kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); Kinase NYK; Protein-tyrosine kinase receptor flk-1; sCD309; soluble CD309; soluble vascular endothelial growth factor receptor 2; soluble VEGFR 2; soluble VEGFR2; tyrosine kinase growth factor receptor; unnamed protein product; Vascular endothelial growth factor receptor 2; vascular endothelial growth factor receptor- 2; vascular endothelial growth factor receptor-2; vascular endothelial growth factor receptor-3; VEGF R2; VEGF receptor-2; VEGFR-2; VEGFR-2; tyrosine kinase

          View more View less

          Gene Aliases: 6130401C07; CD309; Flk-1; FLK1; KDR; Krd-1; Ly73; sVEGFR-2; VEGFR; VEGFR-2; VEGFR2

          View more View less

          UniProt ID: (Human) P35968, (Mouse) P35918

          View more View less

          Entrez Gene ID: (Human) 3791, (Mouse) 16542

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity vascular endothelial growth factor-activated receptor activity integrin binding protein binding ATP binding coreceptor activity kinase activity transferase activity growth factor binding vascular endothelial growth factor binding identical protein binding cadherin binding Hsp90 protein binding transferase activity, transferring phosphorus-containing groups transmembrane signal receptor
          Process(es)
          angiogenesis ovarian follicle development patterning of blood vessels vasculogenesis positive regulation of protein phosphorylation positive regulation of endothelial cell proliferation vasculature development lymph vessel development positive regulation of mesenchymal cell proliferation epithelial cell maturation endocardium development endothelium development transmembrane receptor protein tyrosine kinase signaling pathway positive regulation of cell proliferation regulation of cell shape mesenchymal cell proliferation positive regulation of endothelial cell migration negative regulation of gene expression positive regulation of macroautophagy cell migration peptidyl-tyrosine phosphorylation hemopoiesis cell differentiation lung development positive regulation of cell migration positive regulation of BMP signaling pathway embryonic hemopoiesis cellular response to vascular endothelial growth factor stimulus vascular endothelial growth factor receptor-2 signaling pathway vascular endothelial growth factor signaling pathway negative regulation of apoptotic process surfactant homeostasis regulation of MAPK cascade positive regulation of MAPK cascade negative regulation of neuron apoptotic process positive regulation of blood vessel endothelial cell migration cell fate commitment endothelial cell differentiation positive regulation of angiogenesis protein autophosphorylation vascular endothelial growth factor receptor signaling pathway lung alveolus development post-embryonic camera-type eye morphogenesis branching morphogenesis of an epithelial tube epithelial cell proliferation positive regulation of epithelial cell proliferation positive regulation of positive chemotaxis positive regulation of focal adhesion assembly positive regulation of protein kinase B signaling positive regulation of mitochondrial depolarization calcium ion homeostasis epithelial tube morphogenesis blood vessel endothelial cell differentiation vascular wound healing positive regulation of ERK1 and ERK2 cascade semaphorin-plexin signaling pathway stem cell proliferation positive regulation of cell migration involved in sprouting angiogenesis positive regulation of mitochondrial fission positive regulation of nitric oxide-cGMP mediated signal transduction regulation of hematopoietic progenitor cell differentiation regulation of bone development cellular response to hydrogen sulfide negative regulation of endothelial cell apoptotic process positive regulation of stem cell proliferation positive regulation of endothelial cell chemotaxis positive regulation of vasculogenesis response to hypoxia regulation of endothelial cell proliferation cell migration involved in sprouting angiogenesis endochondral bone growth protein phosphorylation positive regulation of cytosolic calcium ion concentration multicellular organism development male gonad development positive regulation of phosphatidylinositol 3-kinase signaling phosphorylation positive regulation of vascular endothelial growth factor receptor signaling pathway positive regulation of TOR signaling calcium-mediated signaling using intracellular calcium source peptidyl-tyrosine autophosphorylation response to drug regulation of endothelial cell differentiation positive regulation of vasodilation positive regulation of long-term neuronal synaptic plasticity cell maturation neuron projection morphogenesis positive regulation of calcium-mediated signaling positive regulation of nitric-oxide synthase biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline