What are the quenchers in the VetMAX-Gold SIV Detection Kit?
The VetMAX-Gold SIV Detection Kit contains the following quenchers:
- SIV RNA: Eclipse® Q
- Xeno RNA Control: Eclipse® Q
- SIV RNA: Eclipse® Q
- Xeno RNA Control: Eclipse® Q
Search
View additional product information for VetMAX™-Gold SIV Detection Kit - FAQs (4415200)
3 product FAQs found