TaqMan™ Universal Master Mix II, no UNG
Applied Biosystems™

TaqMan™ Universal Master Mix II, no UNG

TaqMan Universal Master Mix II is a convenient mix of components (except primers, probes, template, and water) necessary to performRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
444004910 x 5 mL
44400431 x 1 mL
44400401 x 5 mL
44400472 x 5 mL
44400485 x 5 mL
44400411 x 50 mL
Catalog number 4440049
Price (USD)
5,453.65
Online exclusive
6,060.00
Save 606.35
Each
Quantity:
10 x 5 mL
Price (USD)
5,453.65
Online exclusive
6,060.00
Save 606.35
Each

TaqMan Universal Master Mix II is a convenient mix of components (except primers, probes, template, and water) necessary to perform a real-time polymerase chain reaction (PCR). Use TaqMan Universal Master Mix II to amplify complementary DNA (cDNA) and DNA targets for a variety of applications, including quantitation and genotyping.

This TaqMan Universal Master Mix II is supplied as a 2X concentrate and contains:
• AmpliTaq Gold DNA Polymerase, UP (Ultra Pure)
• dNTPs (with dUTP)
• ROX Passive Reference
• Optimized buffer components

Note: This TaqMan Universal Master Mix II does not contain uracil-N-glycoslyase (UNG). See a complete listing of all TaqMan Universal Master Mix II products, with and without UNG.

TaqMan Universal Master Mix II has been optimized for use with primers and TaqMan probes that have been designed according to Thermo Fisher Scientific guidelines. The master mix can be used with custom TaqMan assays available from our custom assay service or with pre-optimized assays, such as:
• TaqMan gene expression assays
• TaqMan microRNA assays
• TaqMan drug metabolism genotyping assays
• TaqMan SNP genotyping Assays

For RNA quantitation experiments, TaqMan Universal Master Mix II is used in the second step of a two-step reverse transcription/polymerase chain reaction (RT-PCR) protocol. The cDNA template used with the master mix can be generated in a reverse transcription reaction using cDNA synthesis kits available from Thermo Fisher Scientific.

Alternative product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Universal Master Mix II and added additional capabilities for your gene expression analysis.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)2 X
For Use With (Equipment)7300 System, 7500 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineTaqMan
Product TypeUniversal Master Mix II
Purity or Quality GradeUP (Ultra Pure)
Quantity10 x 5 mL
Sample TypeDNA (Genomic), cDNA
Volume50 mL
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, Genotyping, miRNA analysis
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C in the dark.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

What is the benchtop stability of the TaqMan miRNA Assay mixed with TaqMan Universal Master Mix II (Cat. No. 4440043)?

24 hours.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.