What is the maximum sample volume that can be used when detecting Mycoplasma using the PrepSEQ Mycoplasma Nucleic Acid Extraction kit?
The maximum sample volume is 50 mL (or up to 2 x 10e8 cells).
Search
View additional product information for PrepSEQ™ 1-2-3 Mycoplasma Nucleic Acid Extraction Kit - FAQs (4443789)
1 product FAQs found