PrepSEQ™ 1-2-3 Mycoplasma Nucleic Acid Extraction Kit
Applied Biosystems™

PrepSEQ™ 1-2-3 Mycoplasma Nucleic Acid Extraction Kit

The Applied Biosystems PrepSEQ 1-2-3 Nucleic Acid Extraction Kit rapidly extracts Mycoplasma genomic DNA from 100 μL sample volumes of research cell lines, stem cell lines, and cell and tissue therapy samples.
Have Questions?
Catalog NumberQuantity
4443789100 reactions
Catalog number 4443789
Price (USD)
1,144.00
Each
Quantity:
100 reactions
Price (USD)
1,144.00
Each

The Applied Biosystems PrepSEQ 1-2-3 Nucleic Acid Extraction Kit rapidly extracts Mycoplasma genomic DNA from 100 μL sample volumes of research cell lines, stem cell lines, and cell and tissue therapy samples. In the PrepSEQ 1-2-3 kit protocol, a rapid procedure recovers DNA from total lysate of the test sample using magnetic-bead based technology. The PrepSEQ 1-2-3 kit is optimized for use with the MycoSEQ Mycoplasma Detection Kit.

Features of the PrepSEQ 1-2-3 Nucleic Acid Extraction Kit include:

  • High recovery of Mycoplasma genomic DNA from mammalian cell culture samples
  • Rapid easy-to-use protocol with results in under 2 hours
  • Compatible with the MycoSEQ Mycoplasma Detection assay
  • Protocol available for automated sample preparation

High DNA recovery sample preparation
Optimized for use with the MycoSEQ Mycoplasma Detection Kit, the PrepSEQ 1-2-3 Nucleic Acid Extraction Kit uses proprietary magnetic bead-based separation technology to extract Mycoplasma DNA from mammalian cell lines. The PrepSEQ 1-2-3 kit extracts Mycoplasma genomic DNA from research cell lines, stem cell lines, and cellular and tissue therapy samples with high efficiency.

The protocol is recommended for use with a starting sample volume of 100 μL with a maximum cell density of 106 cells. The PrepSEQ 1-2-3 kit provides an easy to use procedure consisting of 1) RNase treatment, 2) proteinase treatment, and 3) DNA extraction using the magnetic bead technology (figure below).

The kit isolates Mycoplasma genomic DNA with high sensitivity from total lysate of test sample. The PrepSEQ 1-2-3 kit and the MycoSEQ kit allow routine screening of high-value cell lines using an easy, rapid and sensitive protocol. The figure below demonstrates how a sample that was spiked with low levels of Mycoplasma bovoculi DNA was found negative for Mycoplasma contamination using a commercially prepared kit. When using the PrepSEQ 1-2-3 kit and the MycoSeq kit, the correct positive call for Mycoplasma contamination was made.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Final Product TypeGenomic DNA
High-throughput CompatibilityHigh-throughput Compatible
No. of Reactions100 Reactions
Product LinePrepSEQ
Quantity100 reactions
Sample TypeCell Culture Supernatants
Sample Volume100 μL
ScaleMini
Target Organism ClassMycoplasma spp.
Test Time2 hr.
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
Includes buffers, magnetic beads and enzymes for DNA extraction for Mycoplasma DNA extraction

Frequently asked questions (FAQs)

What is the maximum sample volume that can be used when detecting Mycoplasma using the PrepSEQ Mycoplasma Nucleic Acid Extraction kit?

The maximum sample volume is 50 mL (or up to 2 x 10e8 cells).