TaqMan™ Mutation Detection Assay with castPCR™ Technology
Applied Biosystems TaqMan Mutation Detection Assays are powered by competitive allele-specific TaqMan castPCR technology to detect and measure somatic mutations자세히 알아보기
Have Questions?
보기 방식 변경
카탈로그 번호
수량
4465807
Gene Reference (75 reactions), Inventoried
4465804
Mutant Allele (75 reactions), Inventoried
2 개 옵션
카탈로그 번호 4465807
제품 가격(KRW)
1,017,000
온라인 행사
Ends: 31-Dec-2026
1,271,000
할인액 254,000
Each
수량:
Gene Reference (75 reactions), Inventoried
Applied Biosystems TaqMan Mutation Detection Assays are powered by competitive allele-specific TaqMan castPCR technology to detect and measure somatic mutations in genes associated with cancer research. castPCR technology is highly specific and sensitive, and can detect rare amounts of mutated DNA in a sample that contains large amounts of normal, wild-type DNA. These assays are compatible with different sample types, such as cell lines, formalin-fixed paraffin-embedded (FFPE) tissue samples, and fresh-frozen tissue samples.
castPCR technology Competitive, allele-specific TaqMan PCR utilizes an allele-specific primer for mutant allele detection that competes with an MGB blocker oligonucleotide to suppress the wild-type background.
Somatic mutation detection By combining the power of TaqMan and castPCR technologies, TaqMan Mutation Detection Assays provide:
• Higher specificity—designed to block the wild-type and amplify only the mutant • Higher sensitivity—designed to detect somatic mutations down to 1 cancer cell in 1,000 normal cells • Faster workflow—3 hours from sample to result
With 819 assays for 47 known cancer research genes available today like KRAS, BRAF, KIT, JAK2, the TaqMan Mutation Detection Assays provide an innovative tool for your cancer research.
All TaqMan Mutation Detection Assays have undergone extensive testing to ensure verify high sensitivity and specificity.
사양
Fidelity (Taq 대비)10 X
형식Tube
제품라인TaqMan
제품 유형Mutation Detection Assay
수량Gene Reference (75 reactions), Inventoried
배송 조건Room Temperature
부피150 μL
농도10X
검출 방법Primer-probe
형태Frozen
GC-Rich PCR PerformanceHigh
PCR 방법qPCR
반응 속도Fast
Unit SizeEach
구성 및 보관
1 tube containing a 10X mix of unlabeled PCR primers and a TaqMan MGB probe
Store at -15 to -25°C.
자주 묻는 질문(FAQ)
How can I find a mutant control sample for my Mutation Detection experiment?
You can check the COSMIC database to see if a control sample is identified for your mutation. If one is not available, you can have one synthesized (using our GeneArt Gene Synthesis service, for example).
In a Mutation Detection experiment, what is the difference between the wild-type assay and the gene reference assay?
The wild-type assay targets the same site as the mutant assay, but blocks the mutant allele. The reference assay detects the gene of interest at a mutation-free region on the same gene. The gene reference assay will amplify both mutant and wild-type gDNA, while the wild-type assay will only amplify the wild type. Both the reference assay and the wild-type allele assay can be used in the same manner to determine the percent mutation status of a sample. Wild-type allele assays are only available for the initial subset of assays.
Can I use TaqMan SNP Genotyping Assays to detect single-nucleotide variants (SNVs) that are present in less than 5% of a sample?
TaqMan SNP Genotyping Assays are designed to work on equal volumes, so if the mutation is only present at 5%, it is possible that the assay will not detect the variants. Therefore, we do not recommend using these assays to detect somatic mutations. SNP assays by qPCR with TaqMan SNP Genotyping Assays are intended to detect germline mutations.
For somatic mutations, we recommend using digital PCR or castPCR. Find more information about digital PCR and castPCR in the links below: