TaqMan™ Mutation Detection Assay with castPCR™ Technology
Applied Biosystems™

TaqMan™ Mutation Detection Assay with castPCR™ Technology

Applied Biosystems TaqMan Mutation Detection Assays are powered by competitive allele-specific TaqMan castPCR technology to detect and measure somatic mutationsRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4465804Mutant Allele (75 reactions), Inventoried
4465807Gene Reference (75 reactions), Inventoried
Catalog number 4465804
Price (USD)
1,286.65
Online exclusive
1,446.00
Save 159.35
Each
Quantity:
Mutant Allele (75 reactions), Inventoried
Applied Biosystems TaqMan Mutation Detection Assays are powered by competitive allele-specific TaqMan castPCR technology to detect and measure somatic mutations in genes associated with cancer research. castPCR technology is highly specific and sensitive, and can detect rare amounts of mutated DNA in a sample that contains large amounts of normal, wild-type DNA. These assays are compatible with different sample types, such as cell lines, formalin-fixed paraffin-embedded (FFPE) tissue samples, and fresh-frozen tissue samples.

castPCR technology
Competitive, allele-specific TaqMan PCR utilizes an allele-specific primer for mutant allele detection that competes with an MGB blocker oligonucleotide to suppress the wild-type background.

Somatic mutation detection
By combining the power of TaqMan and castPCR technologies, TaqMan Mutation Detection Assays provide:

• Higher specificity—designed to block the wild-type and amplify only the mutant
• Higher sensitivity—designed to detect somatic mutations down to 1 cancer cell in 1,000 normal cells
• Faster workflow—3 hours from sample to result

With 819 assays for 47 known cancer research genes available today like KRAS, BRAF, KIT, JAK2, the TaqMan Mutation Detection Assays provide an innovative tool for your cancer research.

All TaqMan Mutation Detection Assays have undergone extensive testing to ensure verify high sensitivity and specificity.

Specifications
Fidelity (vs. Taq)10 X
FormatTube
Product LineTaqMan
Product TypeMutation Detection Assay
QuantityMutant Allele (75 reactions), Inventoried
Shipping ConditionRoom Temperature
Concentration10X
Detection MethodPrimer-probe
FormFrozen
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
1 tube containing a 10X mix of unlabeled PCR primers, a TaqMan MGB probe, and an MGB oligonucleotide blocker.

Store at -15 to -25°C.

Frequently asked questions (FAQs)

How can I find a mutant control sample for my Mutation Detection experiment?

You can check the COSMIC database to see if a control sample is identified for your mutation. If one is not available, you can have one synthesized (using our GeneArt Gene Synthesis service, for example).

In a Mutation Detection experiment, what is the difference between the wild-type assay and the gene reference assay?

The wild-type assay targets the same site as the mutant assay, but blocks the mutant allele. The reference assay detects the gene of interest at a mutation-free region on the same gene. The gene reference assay will amplify both mutant and wild-type gDNA, while the wild-type assay will only amplify the wild type. Both the reference assay and the wild-type allele assay can be used in the same manner to determine the percent mutation status of a sample. Wild-type allele assays are only available for the initial subset of assays.

Can I use TaqMan SNP Genotyping Assays to detect single-nucleotide variants (SNVs) that are present in less than 5% of a sample?

TaqMan SNP Genotyping Assays are designed to work on equal volumes, so if the mutation is only present at 5%, it is possible that the assay will not detect the variants. Therefore, we do not recommend using these assays to detect somatic mutations. SNP assays by qPCR with TaqMan SNP Genotyping Assays are intended to detect germline mutations.

For somatic mutations, we recommend using digital PCR or castPCR. Find more information about digital PCR and castPCR in the links below:

Digital PCR Assays and Kits

QuantStudio Absolute Q Digital PCR System

Absolute Q 1-Step RT-dPCR Master Mix (4X) (Cat. No. A55146) User Guide

TaqMan Mutation Detection Assay with castPCR Technology (Cat. Nos. 4465804, 4465807)

TaqMan Mutation Detection Assay: Competitive Allele-Specific TaqMan PCR

Find additional tips, troubleshooting help, and resources within our TaqMan Primers and Probes Support Center.