Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IFN gamma Antibodies

          Invitrogen

          IFN gamma Monoclonal Antibody (4S.B3), PerCP-Cyanine5.5, eBioscience™

          Advanced Verification
          85 Published Figures
          79 References
          View all (238) IFN gamma antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite IFN gamma Monoclonal Antibody (4S.B3), PerCP-Cyanine5.5, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (85)
          • Advanced Verification (1)
          IFN gamma Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          IFN gamma Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 87

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IFN gamma Antibody (45-7319-42) in Flow

          Intracellular staining of normal human peripheral blood cells stimulated for 5 hours with PMA/Ionomycin in the presence of Monensin with Anti-Human CD4 APC (Product # 17-0049-42) and Mouse IgG1 K Isotype Control PerCP-Cyanine5-5 (Product # 45-4714-82) (left) or Anti-Human IFN gamma PerCP-Cyanine5-5 (right). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody in Flow Cytometry (Flow)
          IFN gamma Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          45-7319-42

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          5 µL (0.06 µg)/test
          View 79 publications 79 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Recommended Isotype Control

          Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), PerCP-Cyanine5.5, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          4S.B3

          Conjugate

          PerCP-Cyanine5.5 PerCP-Cyanine5.5 PerCP-Cyanine5.5
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 650
          • eFluor 450
          • eFluor 506
          • eFluor 660
          • FITC
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • Request custom conjugation

          Excitation/Emission Max

          489/679 nm View spectra spectra

          Form

          Liquid

          Concentration

          5 µL/Test

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10718246

          Product Specific Information

          Description: The 4S.B3 monoclonal antibody reacts with interferon-gamma (IFN gamma). Human IFN gamma is a 17 kDa factor produced by activated T and NK cells and is an anti-viral and anti-parasitic cytokine. IFN gamma in synergy with other cytokines, such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.

          Applications Reported: This 4S.B3 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

          Applications Tested: This 4S.B3 antibody has been pre-titrated and tested by intracellular staining of stimulated normal human peripheral blood cells. This can be used at 5 µL (0.06 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

          Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: cytokine; H-IFN-g; IFN γ; IFN-gamma; IFN-y; IFNg; IFNγ; immune interferon; Interferon; Interferon y; Interferon γ; Interferonγ; M-IFN-g; R-IFN-g; unnamed protein product

          View more View less

          Gene Aliases: IFG; IFI; IMD69

          View more View less

          UniProt ID: (Human) P01579

          View more View less

          Entrez Gene ID: (Human) 3458

          View more View less

          Function(s)
          cytokine activity interferon-gamma receptor binding protein binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter microglial cell activation positive regulation of cytokine production adaptive immune response macrophage activation involved in immune response apoptotic process immune response humoral immune response cell surface receptor signaling pathway JAK-STAT cascade positive regulation of cell proliferation response to virus positive regulation of autophagy positive regulation of gene expression negative regulation of gene expression positive regulation of epithelial cell migration macrophage differentiation negative regulation of epithelial cell differentiation negative regulation of interleukin-17 production positive regulation of chemokine production positive regulation of interleukin-1 beta production positive regulation of interleukin-12 production positive regulation of interleukin-23 production positive regulation of interleukin-6 production positive regulation of tumor necrosis factor production positive regulation of peptidyl-serine phosphorylation of STAT protein positive regulation of smooth muscle cell apoptotic process Fc-gamma receptor signaling pathway involved in phagocytosis type III interferon-mediated signaling pathway macrophage activation positive regulation of tyrosine phosphorylation of STAT protein positive regulation of MHC class II biosynthetic process positive regulation of nitric oxide biosynthetic process positive regulation of osteoclast differentiation positive regulation of cell adhesion positive regulation of glycolytic process negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter astrocyte activation negative regulation of smooth muscle cell proliferation positive regulation of inflammatory response positive regulation of phagocytosis positive regulation of neurogenesis regulation of insulin secretion positive regulation of membrane protein ectodomain proteolysis interferon-gamma-mediated signaling pathway positive regulation of fructose 1,6-bisphosphate metabolic process positive regulation of vitamin D biosynthetic process extrinsic apoptotic signaling pathway receptor signaling pathway via STAT cellular response to virus neuroinflammatory response negative regulation of beta-amyloid clearance positive regulation of glutamate receptor signaling pathway positive regulation of cellular respiration positive regulation of beta-amyloid formation positive regulation of protein localization to plasma membrane positive regulation of exosomal secretion positive regulation of iron ion import across plasma membrane positive regulation of tumor necrosis factor (ligand) superfamily member 11 production
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-7d6nq:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline