Periodic acid, for analysis
Periodic acid, for analysis
Periodic acid, for analysis
Thermo Scientific Chemicals

Periodic acid, for analysis

CAS: 10450-60-9 | H5IO6 | 227.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 453170250
Price (USD)
69.60
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
69.60
Each
Chemical Identifiers
CAS10450-60-9
IUPAC Nametetrahydroxyiodous acid
Molecular FormulaH5IO6
InChI KeyTWLXDPFBEPBAQB-UHFFFAOYSA-N
SMILESO[I](O)(O)(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals
Titration Iodimetric99.0 to 101.0 %
Residue after ignition=<0.05 %
Insoluble matter=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only