Immuno Standard Modules, PolySorp, black - FAQs
Immuno Standard Modules, PolySorp, black - FAQs
View additional product information for Immuno Standard Modules White and Black - FAQs (437591, 437915, 437702, 7566, 475515, 475523)
1 product FAQs found
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?
Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.