CD206 (MMR) Monoclonal Antibody (19.2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD206 (MMR) Monoclonal Antibody (19.2), eFluor™ 450, eBioscience™ - FAQs (48-2069-42, 48-2069-41)