Sequential Protein/DNA/RNA Extraction Kit, 24 rxns - FAQs
Sequential Protein/DNA/RNA Extraction Kit, 24 rxns - FAQs
View additional product information for Sequential Protein/DNA/RNA Extraction Kit - FAQs (50000D, 50010D)
25 product FAQs found
How can I get my protein sample ready for LC-MS analysis?
The protein fraction isolated with the Sequential Protein/DNA/RNA Extraction Kit is compatible with LC-MS analysis. In the application note, the protein fraction was prepared using the EasyPep MS Sample Prep Kit, with the lysis step omitted as protein extraction had already been completed. Alternatively, users may prepare the protein fraction using a standard bottom-up proteomics workflow, including protein cleanup or buffer exchange if needed, reduction, alkylation, enzymatic digestion, peptide cleanup/desalting, and LC-MS analysis according to their laboratory’s established protocols.
How compatible is the kit with different lysis buffers or additives (e.g., protease inhibitors, RNase inhibitors)?
The lysis buffer included in the kit has been specifically optimized for efficient lysis of cells and tissue samples and should not be replaced with another lysis buffer. However, it can be supplemented with protease inhibitors. RNase inhibitors are not necessary, as RNA integrity is well maintained.
What are the carry-over rates between analyte fractions (protein–DNA–RNA)?
The protein, DNA and RNA eluates exhibit high purity with very low carry-over between fractions.
Could the kit be used on exosomes and on serum or plasma samples?
Yes, the kit can be used with a plethora of sample types, provided the sample is sufficiently lysed.
Is there a protocol for on-bead digestion for subsequent proteomics analysis?
There is no available protocol for on-bead digestion, although the kit is theoretically compatible with this process. The kit can be combined with the EasyPep MS Sample Prep Kit by skipping the lysis step.
What is the size and binding capacity of the beads?
Dynabeads Sequential Protein Binding Beads have a diameter of 2.8 µm, while MagMAX Sequential DNA/RNA Binding Beads have a diameter of 1 µm. The binding capacity cannot be disclosed here, as it involves proprietary information regarding the binding principle.
Can the remaining lysed samples be kept for later use?
Yes, the remaining lysate from the sequential workflow can be stored at -80°C and used later without impacting the yield and quality of the analytes.
Can I expect similar yields with this kit versus standard bead-based or column-based kits for single isolation of DNA, RNA, and protein?
Yes, the yields obtained using the kit are comparable to those obtained with single analyte isolation kits. However, the yields of proteins are usually slightly lower than cells lysed directly with RIPA buffer.
As RNA is the last analyte to be isolated, is RNA integrity good enough for downstream analysis?
The buffers and protocols have been optimized to help ensure good RNA integrity.
What is the advantage of the kit over single analyte extraction kits?
The kit enables high sensitivity from small sample input sizes, making it exceptional for precious samples (e.g., patient samples) or samples that cannot be split for separate extraction of protein, DNA, and RNA (e.g., heterogeneous tissue specimens).
What is the duration of the entire workflow?
The manual protocol uses ~2 hours of hands-on time for 10 samples. The automated protocol uses ~1.5 hours of instrument run time and 30–60 minutes of hands-on time for 12 to 96 samples.
Is the kit compatible with liquid handlers?
The kit is magnetic bead-based and thus compatible with liquid handlers (e.g., Hamilton NIMBUS Presto System).
What are the advantages of the kit over column-based extraction?
The kit uses magnetic beads for the specific isolation of protein, DNA, and RNA, which enables automation of the workflow, resulting in higher reproducibility and throughput. Additionally, our optimized buffer chemistry increases sensitivity, allowing the use ofsmaller sample sizes from the start. Lastly, the protein can be directly prepared for mass spectrometry analysis without further desalting.
Is the kit compatible with blood samples?
The kit is developed for cells and is not directly compatible with whole blood samples. We recommend pre-isolating cells from blood, such as peripheral blood mononuclear cells (PBMCs), prior to protein, DNA, and RNA extraction for optimal results.
Is the kit compatible with fungi, plant cells, or yeast?
While these sample types have not been specifically tested with our kit, theoretically they should work, provided the sample is sufficiently lysed. Proper lysis is crucial to help ensure effective isolation of protein, DNA, and RNA from fungi, plant cells, or yeast.
What kind of sequencing methods are compatible?
The kit is compatible with targeted sequencing (e.g., Ion Torrent next-generation-sequencing (NGS) systems and short-read sequencing (e.g., Illumina NGS platforms).
Is the kit compatible with long-read sequencing technologies?
The kit has not been assessed for long-read sequencing.
Is the Thermo Scientific Sequential Protein/DNA/RNA Extraction Kit compatible with the Olink Proximity Extension Assay (PEA) technology?
Preliminary proof-of-concept data suggest that the Sequential Protein/DNA/RNA Extraction Kit is compatible with Olink PEA-based proteomic analysis. Depending on the purpose of the analysis and sample type, further optimization of the sample preparation workflow may be required.
Can I use the kit with formalin-fixed, paraffin-embedded (FFPE) samples?
The kit has not been optimized for use with FFPE samples. Additionally, FFPE samples are not ideal for RNA integrity, which may affect the quality and yield of RNA isolated from these samples. For optimal results, we recommend using fresh or frozen tissue samples.
Can I isolate microRNAs with the kit?
Yes, this kit is designed to isolate total RNA, which includes small RNAs like microRNAs.
What is the size range of the proteins that are isolated from the kit? Can I isolate peptides?
The size of the isolated proteins ranges from around 5 to 590 kDa. The isolation of specific peptides has not been assessed.
Is it possible to use a sample of cells or tissue that is already in a buffer? (e.g., sorted cells in PBS)
Yes, the lysis buffer’s effectiveness has been tested by diluting it up to 2 times, with performance like the non-diluted lysis buffer.
What is the maximal number of cells or tissue mass that can be used per extraction?
The kit is optimized for a maximum input of 1,000,000 cells or 10 mg of tissue per extraction.
What is the minimal number of cells or tissue mass that can be used in the workflow?
The kit is optimized for 100,000 cells and 1 mg of tissue. We have tested down to 1,000 cells and 0.005 mg of tissue, demonstrating linearity in the yields of protein, DNA, and RNA.
Can I pause the workflow and freeze my sample after protein/DNA isolation?
Yes, the sequential workflow can be paused after protein isolation and after DNA isolation by freezing the sample plate at -20°C, without impacting the yield and quality of the analytes.