Sequential Protein/DNA/RNA Extraction Kit
Thermo scientific™

Sequential Protein/DNA/RNA Extraction Kit

The Sequential Protein/DNA/RNA Extraction Kit represents an exceptional multi-omics solution designed for the sequential purification of high-quality protein, DNA, and RNA from the same cell or tissue sample. This bead-based extraction system addresses the critical need for comprehensive molecular analysis when sample availability is limited or when studying heterogeneous specimens.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
50000D24 Reactions
50010D100 Reactions
Catalog number 50000D
Price (USD)
440.00
Each
No. of Reactions:
24 Reactions
Price (USD)
440.00
Each

The Sequential Protein/DNA/RNA Extraction Kit represents an exceptional multi-omics solution designed for the sequential purification of high-quality protein, DNA, and RNA from the same cell or tissue sample. This bead-based extraction system addresses the critical need for comprehensive molecular analysis when sample availability is limited or when studying heterogeneous specimens.

The product is available as a 24 reaction kit (Cat. No. 50000D) and a 100 reaction kit (50010D). The 100-reaction vials of beads, buffers, and solutions are also available separately.

Features of the Sequential Protein/DNA/RNA Extraction Kit include:

  • High sensitivity—supports scalable extractions from precious or low sample volumes down to 1,000 cells or 0.005 mg tissue (tissue dependent)
  • Direct compatibility—extracted analytes down to 100,000 cells or 1-2 mg tissue are validated and suitable for mass spectrometry and NGS applications
  • Automated high-throughput—compatible with KingFisher instruments for scalable processing
  • Enhanced reproducibility—standardized protocol reduces variability

The kit employs a bead-based sequential extraction methodology utilizing protein isolation prior to nucleic acid extraction, resulting in enhanced protein yields that are directly compatible with mass spectrometry analysis. This approach enables researchers to obtain analytes suitable for diverse downstream applications including SDS-PAGE, western blotting, mass spectrometry, sequencing, genotyping, and qPCR from sample inputs as small as 1-2 mg of tissue or 100,000 cells while maintaining quality standards required for both next-generation sequencing (NGS) and MS platforms. The Dynabeads and MagMAX magnetic bead-based purification technology helps enable high reproducibility and scalable, high-throughput processing when integrated with automation systems such as the KingFisher Sample Purification System.

This kit combines the advantages of multi-analyte extraction with the precision and efficiency required for advanced proteomics and genomics workflows, making it particularly valuable for multi-omics studies where sample conservation and analytical comprehensiveness are paramount.

Verified performance

The kit helps deliver yield and quality metrics comparable to standard single-analyte extraction kits, facilitating efficient multi-analyte recovery from a single sample. While most commercial sequential protocols are optimized for large sample volumes (e.g. 1 million cells or 10–20 mg tissue), this kit offers exceptional versatility across a wider range. Validated for 100,000 cells and 1–2 mg tissue, it has also been successfully used with as few as 1,000 cells and 5 μg tissue while maintaining linear yields and preserving nucleic acid integrity suitable for qPCR analysis. This broad dynamic range provides greater flexibility and enables efficient processing of precious or limited samples that are difficult to analyze with conventional methods.

Streamlined three-step sequential extraction

The extraction procedure follows a systematic three-step process designed to support a high recovery of each biomolecule class. Initially, Dynabeads Protein Beads are introduced to the lysed sample and incubated to facilitate protein binding, after which they are transferred to a separate tube for washing and protein processing compatible with downstream western blot or mass-spectrometry applications. Subsequently, MagMAX DNA/RNA Binding beads are added to the original tube, now depleted of proteins, to enable DNA binding and extraction using proprietary buffer formulations before transferring to a new tube for further preparation. Finally, MagMAX DNA/RNA Binding beads are again introduced to the original tube, now depleted of both proteins and DNA, to facilitate RNA binding and extraction using specialized proprietary buffers and solutions, with subsequent transfer to a dedicated tube for final preparation. The resulting extracted proteins are suitable for WB or MS analysis, while the recovered DNA and RNA are compatible with quantitative PCR (qPCR) and next-generation sequencing (NGS) applications.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bead TypeMagnetic Polystyrene
For Use With (Application)Mass Spectrometry, Western Blot, qPCR, Sequencing
For Use With (Equipment)KingFisher™ Duo Prime, Flex, or Apex Purification Systems
FormulationReagent Solution
No. of Reactions24 Reactions
Product LineMagMAX, Dynabeads
Product TypeMagnetic Bead Protein Purification System
Quantity1 Kit
Regulatory StatusFor Research Use Only
Sample TypeCell Lines, PBMC, Fresh/Frozen Tissue
Shipping ConditionAmbient Temperature
Size1 to 3 μm
ThroughputFlexible
FormatKit
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
•The Kit contains 2 vials of magnetic beads and the 10 buffers/solutions required to extract the protein, DNA and RNA from the sample.
•Store the Dynabeads Sequential Protein Binding Beads and the Sequential Protein Elution Buffer at 2°C to 8°C. Store the other components of the kit at 15°C to 25°C.

Frequently asked questions (FAQs)

How can I get my protein sample ready for LC-MS analysis?

The protein fraction isolated with the Sequential Protein/DNA/RNA Extraction Kit is compatible with LC-MS analysis. In the application note, the protein fraction was prepared using the EasyPep MS Sample Prep Kit, with the lysis step omitted as protein extraction had already been completed. Alternatively, users may prepare the protein fraction using a standard bottom-up proteomics workflow, including protein cleanup or buffer exchange if needed, reduction, alkylation, enzymatic digestion, peptide cleanup/desalting, and LC-MS analysis according to their laboratory’s established protocols.

How compatible is the kit with different lysis buffers or additives (e.g., protease inhibitors, RNase inhibitors)?

The lysis buffer included in the kit has been specifically optimized for efficient lysis of cells and tissue samples and should not be replaced with another lysis buffer. However, it can be supplemented with protease inhibitors. RNase inhibitors are not necessary, as RNA integrity is well maintained.

What are the carry-over rates between analyte fractions (protein–DNA–RNA)?

The protein, DNA and RNA eluates exhibit high purity with very low carry-over between fractions.

Could the kit be used on exosomes and on serum or plasma samples?

Yes, the kit can be used with a plethora of sample types, provided the sample is sufficiently lysed.

Is there a protocol for on-bead digestion for subsequent proteomics analysis?

There is no available protocol for on-bead digestion, although the kit is theoretically compatible with this process. The kit can be combined with the EasyPep MS Sample Prep Kit by skipping the lysis step.